Skip to Content
Merck
All Photos(1)

Key Documents

35848

Sigma-Aldrich

2′,7′-Dichlorofluorescein

BioReagent, suitable for fluorescence, ≥90% (T)

Synonym(s):

DCF, Dichlorofluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10Cl2O5
CAS Number:
Molecular Weight:
401.20
Beilstein:
58009
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥90% (T)

form

solid

mp

280 °C (dec.) (lit.)

fluorescence

λex 504 nm; λem 529 nm in 0.1 M Tris pH 8.0

suitability

suitable for fluorescence

SMILES string

Oc1cc2Oc3cc(O)c(Cl)cc3C4(OC(=O)c5ccccc45)c2cc1Cl

InChI

1S/C20H10Cl2O5/c21-13-5-11-17(7-15(13)23)26-18-8-16(24)14(22)6-12(18)20(11)10-4-2-1-3-9(10)19(25)27-20/h1-8,23-24H

InChI key

VFNKZQNIXUFLBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2′,7′-Dichlorofluorescein, also known as DCF is a fluorescent dye. The DCF fluorescence excitation is at 498nm with emission at 522nm. The lipophilic non-fluorescent Dcfh-DA readily crosses the cell membrane through passive diffusion followed by deacetylation. The deacylated product is an oxidant sensitive 2′,7′-dichlorofluorescein (DCHF). DCHF is oxidized later to form highly fluorescent DCF, which is measured.

Application

2′,7′-Dichlorofluorescein is used as fluorescent probe to measure ROS (Reactive Oxygen Species) for biological studies. It may also be used as a dip reagent for the fluorometric determination of vicinal diols, glycosides, and phenols.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W.D. Pfeffer et al.
Journal of Chromatography A, 506, 401-401 (1990)
H. Watanabe et al.
Analytical Sciences, 2, 461-461 (1986)
C Rota et al.
Free radical biology & medicine, 27(7-8), 873-881 (1999-10-09)
The oxidation of 2'-7'-dichlorofluorescin (DCFH) to the fluorescent 2'-7'-dichlorofluorescein (DCF) by horseradish peroxidase (HRP) was investigated by fluorescence, absorption, and electron spin resonance spectroscopy (ESR). As has been previously reported, HRP/H2O2 oxidized DCFH to the highly fluorescent DCF. However, DCF
Masahito Miura et al.
Journal of molecular and cellular cardiology, 56, 97-105 (2012-12-26)
Ca(2+) waves are initiated not only by Ca(2+) leak from the sarcoplasmic reticulum (SR), but also by Ca(2+) dissociation from the myofilaments in the myocardium with nonuniform contraction. We investigated whether contractile properties and the production of reactive oxygen species
Xiuping Chen et al.
Free radical research, 44(6), 587-604 (2010-04-08)
Reactive oxygen species (ROS) are critically important chemical intermediates in biological studies, due to their multiple physiologically essential functions and their often pathologically deleterious effects. Consequently, it is vital that their presence in biological samples has to be quantifiable. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service