Skip to Content
Merck
All Photos(4)

Key Documents

410217

Sigma-Aldrich

2′,7′-Dichlorofluorescein

ACS reagent

Synonym(s):

Dichlorofluorescein; Fluorescein 27

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10Cl2O5
CAS Number:
Molecular Weight:
401.20
Beilstein:
58009
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

form

powder

technique(s)

titration: suitable

color

orange to red-brown, powder

mp

280 °C (dec.) (lit.)

solubility

alcohol: passes test

density

0.790 g/cm3

λmax

509 nm

suitability

passes test for adsorption indicator

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1cc2Oc3cc(O)c(Cl)cc3C4(OC(=O)c5ccccc45)c2cc1Cl

InChI

1S/C20H10Cl2O5/c21-13-5-11-17(7-15(13)23)26-18-8-16(24)14(22)6-12(18)20(11)10-4-2-1-3-9(10)19(25)27-20/h1-8,23-24H

InChI key

VFNKZQNIXUFLBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2′,7′-Dichlorofluorescein is an oxidation-sensitive fluorescent probe.

Application

2′,7′-Dichlorofluorescein has been used to measure ROS (reactive oxygen species) formation in cells. It has been used as a fluorogenic substrate to study the peroxidase activity of saliva samples and extracts obtained from submandibular glands.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andréa G K Ferreira et al.
Journal of cellular biochemistry, 113(1), 174-183 (2011-09-02)
The present study investigated the effects of chronic hyperprolinemia on oxidative and metabolic status in liver and serum of rats. Wistar rats received daily subcutaneous injections of proline from their 6th to 28th day of life. Twelve hours after the
L Foucaud et al.
Toxicology in vitro : an international journal published in association with BIBRA, 24(6), 1512-1520 (2010-07-20)
The aim of this study was to investigate whether carbon black (CB) nanoparticles might induce toxicity to monocytic cells in vitro via an oxidative stress mechanism involving formation of the lipid peroxidation product 4-hydroxynonenal (4-HNE) and the subsequent role of
Olga N Pakhomova et al.
Archives of biochemistry and biophysics, 527(1), 55-64 (2012-08-23)
Nanosecond pulsed electric field (nsPEF) is a novel modality for permeabilization of membranous structures and intracellular delivery of xenobiotics. We hypothesized that oxidative effects of nsPEF could be a separate primary mechanism responsible for bioeffects. ROS production in cultured cells
Janaína Kolling et al.
Cardiovascular toxicology, 11(1), 67-73 (2010-11-16)
Hyperhomocysteinemia is a risk factor for cardiovascular disease, stroke, and thrombosis; however, the mechanisms by which homocysteine triggers these dysfunctions are not fully understood. In the present study, we investigated the effect of chronic hyperhomocysteinemia on some parameters of oxidative
Aleksey Rukavishnikov et al.
Analytical biochemistry, 419(1), 9-16 (2011-08-27)
Cephalosporin was used to synthesize soluble and precipitating fluorogenic β-lactam substrates that demonstrated differential catalytic hydrolysis by three different subtypes of β-lactamase: TEM-1 (class A), p99 (class C), and a Bacillus cereus enzyme sold by Genzyme (class B). The most

Questions

1–4 of 4 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What excitation /emission wavelengths are best for fluorescence detection of 2´,7´-Dichlorofluorescein?

    1 answer
    1. 2´,7´-Dichlorofluorescein can be detected at an excitation wavelength at 504 nm and emission wavelength at 529 nm in 0.1 M Tris pH 8.0.

      Helpful?

  3. In what types of applications can 2´,7´-Dichlorofluorescein be used?

    1 answer
    1. Some applications for use of 2´,7´-Dichlorofluorescein are listed below. This product can be used for the determination of carbohydrates by fluorescence densitometry after TLC.  See G. Gübitz et al., J. Chromatogr., 117, 337, (1976). 2´,7´-Dichlorofluorescein as also been used in the determination of H2O2 by chemiluminescence. See the following references by H. Watanabe et al. Anal. Sci. 2, 461, (1986) and T. Segawa et al. Anal. Sci. 6, 763, (1990). This product has also been used as a  fluorescent additive used in indirect fluorimetric detection in OTC-HPLC. See W.D. Pfeffer and E.S. Yeung, J. Chromatogr., 506, 401, (1990).

      Helpful?

  4. What can be used to make a solution of 2´,7´-dichlorofluorescein?

    1 answer
    1. 2´,7´-Dichlorofluorescein is soluble at 25 mg/mL absolute ethanol with heating. 2´,7´-Dichlorofluorescein is also soluble in water at 0.1 mg/mL.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service