Skip to Content
Merck
All Photos(1)

Key Documents

D4505

Sigma-Aldrich

5,5-Diphenylhydantoin sodium salt

≥99%

Synonym(s):

5,5-Diphenyl-2,4-imidazolidinedione, Phenytoin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C15H11N2O2Na
CAS Number:
Molecular Weight:
274.25
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥99%

form

powder

solubility

aqueous base: soluble

SMILES string

[Na]N1C(=O)NC(C1=O)(c2ccccc2)c3ccccc3

InChI

1S/C15H12N2O2.Na/c18-13-15(17-14(19)16-13,11-7-3-1-4-8-11)12-9-5-2-6-10-12;/h1-10H,(H2,16,17,18,19);/q;+1/p-1

InChI key

FJPYVLNWWICYDW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

5,5-Diphenylhydantoin sodium salt is a derivative of phenytoin. 5,5-Diphenylhydantoin is used for treating epilepsy and has antiepileptic activity. It functions by blocking the sodium channels and modulates excitation of neurons. However, the side effects include hypertrichosis and overgrowth of gum tissue around the teeth. It promotes proliferation of the neural stem cells by modulating growth factors. Diphenylhydantoin is toxic and induces apoptosis in cultured cerebellar granule neurons.

Application

5,5-Diphenylhydantoin sodium salt has been used:
  • in the cream preparation for treating burn wounds
  • in seizure behavior testing as an anticonvulsant in Drosophila
  • in in vivo embryo toxicity test in mouse embryonic stem cells

Biochem/physiol Actions

Reduces incidence of grand mal seizures; appears to stabilize excitable membranes perhaps through effects on Na+, K+, and Ca2+ channels.
Reduces incidence of grand mal seizures; appears to stabilize excitable membranes.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of efficacy of topical phenytoin with hypericin in second-degree burn wound healing: an experimental study in rats
Sayar H, et al.
Medical Science Monitor Basic Research, 20(6), 36-36 (2014)
Diphenylhydantoin promotes proliferation in the subventricular zone and dentate gyrus
Galvez-Contreras AY, et al.
American journal of neuroscience, 3(1), 1-1 (2012)
Phenytoin: a step by step insight into its multiple mechanisms of action?80 years of mechanistic studies in neuropharmacology
HJM Keppel
Journal of Neurology, 264(9), 2043-2047 (2017)
Jinsil Park et al.
The Cleft palate-craniofacial journal : official publication of the American Cleft Palate-Craniofacial Association, 58(6), 697-706 (2021-05-29)
Cleft palate is among the most frequent congenital defects in humans. While gene-environment multifactorial threshold models have been proposed to explain this cleft palate formation, only a few experimental models have verified this theory. This study aimed to clarify whether
An RNAi-mediated screen identifies novel targets for next-generation antiepileptic drugs based on increased expression of the homeostatic regulator pumilio
Lin WH, et al.
Journal of Neurogenetics, 32(2), 106-117 (2018)

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service