Skip to Content
Merck
All Photos(1)

Key Documents

1535008

USP

Phenytoin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5,5-Diphenylhydantoin, 5,5-Diphenyl-2,4-imidazolidinedione, Phenytoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27
Beilstein:
384532
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

phenytoin

manufacturer/tradename

USP

mp

293-295 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=C1NC(=O)C(N1)(c2ccccc2)c3ccccc3

InChI

1S/C15H12N2O2/c18-13-15(17-14(19)16-13,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H,(H2,16,17,18,19)

InChI key

CXOFVDLJLONNDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phenytoin USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Phenytoin Sodium
  • Phenytoin Chewable Tablets
  • Phenytoin Oral Suspension
  • Phenytoin Sodium Injection
  • Extended Phenytoin Sodium Capsules
  • Phenytoin Compounded Topical Gel
  • Fosphenytoin Sodium Injection

Biochem/physiol Actions

Anticonvulsant.
Reduces incidence of grand mal seizures; appears to stabilize excitable membranes perhaps through effects on Na+, K+, and Ca2+ channels.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Disclaimer

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deborah Pugin et al.
Critical care (London, England), 18(3), R103-R103 (2014-06-03)
Seizures refractory to third-line therapy are also labeled super-refractory status epilepticus (SRSE). These seizures are extremely difficult to control and associated with poor outcome. We aimed to characterize efficacy and side-effects of continuous infusions of pentobarbital (cIV-PTB) treating SRSE. We
H Kamp et al.
Bioanalysis, 4(18), 2291-2301 (2012-10-11)
BASF and Metanomics have built-up the database MetaMap(®)-Tox containing rat plasma metabolome data for more than 500 reference compounds. Phenytoin was administered to five Wistar rats of both sexes at dietary dose levels of 600 and 2400 ppm over 28
Fraser Birse et al.
The Cochrane database of systematic reviews, 5(5), CD009485-CD009485 (2012-05-18)
Antiepileptic drugs have been used in pain management since the 1960s; some have shown efficacy in treating different neuropathic pain conditions. Phenytoin is an established antiepileptic drug that has been used occasionally to treat intractable trigeminal neuralgia. To assess the
Anoop S V Shah et al.
Clinical toxicology (Philadelphia, Pa.), 48(8), 800-805 (2010-10-07)
Seizures are a common sequela of self-poisoning. However, their mechanism differs from seizures of other etiologies. Toxicological seizures result from alterations in the excitatory and inhibitory balance of otherwise normal neurons. In contrast, idiopathic or trauma related seizures usually start
Precision, bias, and clinical utility of the Sheiner-Tozer equation to guide phenytoin dosing in critically ill adults.
Jennifer Bolt et al.
Journal of clinical pharmacology, 53(4), 451-455 (2013-02-26)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service