Skip to Content
Merck
All Photos(1)

Documents

239658

Supelco

Bisphenol A

≥99%

Synonym(s):

2,2-Bis(4-hydroxyphenyl)propane, 4,4′-Isopropylidenediphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(C6H4OH)2
CAS Number:
Molecular Weight:
228.29
Beilstein:
1107700
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Assay

≥99%

form

solid

bp

220 °C/4 mmHg (lit.)

mp

158-159 °C (lit.)

SMILES string

CC(C)(c1ccc(O)cc1)c2ccc(O)cc2

InChI

1S/C15H16O2/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12/h3-10,16-17H,1-2H3

InChI key

IISBACLAFKSPIT-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

General description

Bisphenol A (BPA) is a diphenylmethane derivative, commonly used in the production of polycarbonate plastics, epoxy resins, and various other plastic-based consumer products. It is also known to exhibit strong endocrine-disrupting ability.

Application

The analytical standard can also be used as follows:
  • Quantitative analysis of bisphenol A and bisphenol S in three plastic product samples of a mineral water bottle, polycarbonate bottle, and baby bottle, using a modified glassy carbon electrode (GCE) sensor based on a crystalline covalent organic framework (COF)
  • Simultaneous determination of bisphenol A, S, and F in 112 samples of thermal paper receipts by liquid chromatography coupled to mass spectrometry (LC-MS) following their ultrasound-assisted extraction
  • Development and validation of a gas chromatography-tandem mass spectrometry (GC-MS/MS) method for the measurement of bisphenol A and 18 BPA-related compounds in 30 breast milk samples after their solid phase extraction (SPE)
  • Construction of an electrochemical sensor based on multi-walled carbon nanotubes (MWCNTs) modified glassy carbon electrode (GCE) for the determination of bisphenol A & S in river water and thermal paper samples by cyclic voltammetry (CV)
  • Determination of bisphenol A in urine samples using a novel method based on miniaturized QuEChERS extraction and gas chromatography-mass spectrometry (GC-MS)
  • Investigate the bisphenol A content from glass or Pasteur pipettes in biological liquids from maternal women using high-performance liquid chromatography (HPLC).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

440.6 °F - closed cup

Flash Point(C)

227 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjay Basak et al.
Reproductive toxicology (Elmsford, N.Y.), 82, 72-79 (2018-10-24)
Humans are exposed to Bisphenol A (BPA) from the consumer products and plastic substances. However, impacts of low levels of BPA exposure on placental developmental processes such as first trimester trophoblast cell growth, angiogenesis and epigenetic modifications are not well
Maternal bisphenol-A levels at delivery: a looming problem?
Padmanabhan V, et al.
Journal of Perinatology : Official Journal of the California Perinatal Association, 28, 258-258 (2008)
Albert A Koelmans et al.
Environmental pollution (Barking, Essex : 1987), 187, 49-54 (2014-01-21)
It is often assumed that ingestion of microplastics by aquatic species leads to increased exposure to plastic additives. However, experimental data or model based evidence is lacking. Here we assess the potential of leaching of nonylphenol (NP) and bisphenol A
R E FitzGerald et al.
Toxicology letters, 230(2), 368-374 (2014-05-17)
Bisphenol A (BPA) is one of the most widely used and extensively studied chemicals. Numerous studies have reported in vitro effects or animal adverse findings at BPA doses lower than the no observed adverse effect levels (NOAELs) established in regulatory
Tinne Geens et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(10), 3725-3740 (2012-08-15)
Due to the large number of applications of bisphenol-A (BPA), the human exposure routes are multiple. We aimed to review shortly the food and non-food sources of BPA, and to evaluate their contribution to the human exposure. Food sources discussed

Protocols

A comprehensive sample preparation and analytical procedure was developed by Supelco for determining BPA in drinking water.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service