Skip to Content
Merck
All Photos(3)

Key Documents

D4007

Sigma-Aldrich

5,5-Diphenylhydantoin

≥98%

Synonym(s):

5,5-Diphenyl-2,4-imidazolidinedione, Phenytoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27
Beilstein:
384532
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98%

form

powder

mp

293-295 °C (lit.)

solubility

DMSO: soluble

SMILES string

O=C1NC(=O)C(N1)(c2ccccc2)c3ccccc3

InChI

1S/C15H12N2O2/c18-13-15(17-14(19)16-13,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H,(H2,16,17,18,19)

InChI key

CXOFVDLJLONNDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5,5-Diphenylhydantoin is an anticonvulsant, which is used to treat tonic-clonic seizures, complex partial seizures and neurosurgical related seizures. It induces osteoblast proliferation and differentiation.[1] 5,5-Diphenylhydantoin used to treat epilepsy.[2] It reduces the hyperexcitability of tissues and may be associated with cerebellar atrophy.[1]

Application

5,5-Diphenylhydantoin has been used for phenytoin treatment.[3] It has also been used to slow down or prevent mesoendoderm cell migration.[4]

Biochem/physiol Actions

Anticonvulsant.
Reduces incidence of grand mal seizures; appears to stabilize excitable membranes perhaps through effects on Na+, K+, and Ca2+ channels.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

In Vitro Micropatterned Human Pluripotent Stem Cell Test (muP-hPST) for Morphometric-Based Teratogen Screening
Xing J, et al.
Scientific Reports, 7(1), 8491-8491 (2017)
Neuroimaging Pharmacopoeia (2015)
Phenytoin activates SMAD3 phosphorylation and periostin expression in drug-induced gingival enlargement
Kim SS, et al.
Histology and Histopathology, 33(12), 1287-1298 (2018)
Advanced Therapy in Epilepsy (2009)
Jose Brea et al.
SLAS discovery : advancing life sciences R & D, 29(3), 100149-100149 (2024-03-17)
The purpose of the protocol reported in this work is the solubility profiling of large chemical libraries using nephelometry. This technique allows the qualitative classification of compounds as highly, moderately, or poorly water-soluble. The described methodology is not intended to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service