Skip to Content
Merck
All Photos(1)

Key Documents

D184

Sigma-Aldrich

Diethylamine NONOate sodium salt hydrate

≥97% (NMR), crystalline, nitrix oxide donor

Synonym(s):

1,1-Diethyl-2-hydroxy-2-nitroso-hydrazine sodium, 2-(N,N-Diethylamino)-diazenolate 2-oxide sodium salt hydrate, DEA NONOate sodium salt hydrate, Diethylamine/nitric oxide complex sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2N–N(N=O)O- Na+ xH2O
CAS Number:
Molecular Weight:
155.13 (anhydrous basis)
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Diethylamine NONOate sodium salt hydrate, ≥97% (NMR)

Quality Level

Assay

≥97% (NMR)

form

crystalline

storage condition

desiccated
protect from light

color

white

mp

45 °C (lit.)

solubility

H2O: >10 mg/mL
methanol: >25 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

O.[Na+].CCN(CC)N([O-])N=O

InChI

1S/C4H10N3O2.Na.H2O/c1-3-6(4-2)7(9)5-8;;/h3-4H2,1-2H3;;1H2/q-1;+1;

InChI key

BVCZSYKIXSAMOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diethylamine NONOate sodium salt hydrate has been used as a nitric oxide donor to evaluate endothelium-independent dilation.[1][2] It has also been used to determine the level of nitric oxide in cell free systems.[3]

Biochem/physiol Actions

Diethylamine NONOate is a complex of diethylamine with nitric oxide used to generate a controlled release of nitric oxide in solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nina Jeppesen Edin et al.
Journal of radiation research, 54(6), 1015-1028 (2013-05-21)
In this study, a mechanism in which low-dose hyper-radiosensitivity (HRS) is permanently removed, induced by low-dose-rate (LDR) (0.2-0.3 Gy/h for 1 h) but not by high-dose-rate priming (0.3 Gy at 40 Gy/h) was investigated. One HRS-negative cell line (NHIK 3025)
The functional and structural changes in the basilar artery due to overpressure blast injury
Toklu HZ, et al.
Journal of Cerebral Blood Flow and Metabolism, 35(12), 1950-1956 (2015)
High dietary fructose does not exacerbate the detrimental consequences of high fat diet on basilar artery function
Toklu HZ, et al.
Journal of Physiology And Pharmacology, 67(2), 205-205 (2016)
Francesco Tamagnini et al.
The Journal of physiology, 591(16), 3963-3979 (2013-05-15)
Synaptic plasticity in perirhinal cortex is essential for recognition memory. Nitric oxide and endocannabinoids (eCBs), which are produced in the postsynaptic cell and act on the presynaptic terminal, are implicated in mechanisms of long-term potentiation (LTP) and long-term depression (LTD)
Extracellular Alpha-Synuclein Oligomers Induce Parkin S-Nitrosylation: Relevance to Sporadic Parkinson?s Disease Etiopathology
Wilkaniec A, et al.
Molecular Neurobiology, 56(1), 125-140 (2019)

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service