Skip to Content
Merck
All Photos(2)

Key Documents

L2020

Sigma-Aldrich

Mouse Laminin

Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane
1 of 4 reviewers received a sample product or took part in a promotion

from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 0.2 μm filtered, liquid, 1-2 mg/mL, suitable for cell culture

Synonym(s):

Laminin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75
Pricing and availability is not currently available.

Product Name

Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture

biological source

mouse (Engelbreth-Holm-Swarm mouse sarcoma basement membrane)

Quality Level

product line

BioReagent

form

aqueous solution

mol wt

A subunit 400 kDa
B1 subunit 210 kDa
B2 subunit 200 kDa

packaging

pkg of 1 mg

concentration

1-2 mg/mL in Tris-buffered saline

technique(s)

cell culture | mammalian: suitable

surface coverage

1‑2 μg/cm2

impurities

Microbial Contamination, passes test

NCBI accession no.

UniProt accession no.

Binding Specificity

Peptide Source: Collagen

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Laminins are non-collagenous adhesive glycoproteins[1] with a substantial molecular weight. They are heterotrimeric proteins with three disulfide-linked polypeptides known as the α, β, and γ chains.[2] The multiple polypeptide domains of laminins can bind to multiple cell surface receptors, including several integrin dimers, dystroglycan and syndecans, and other ECM components. This extracellular matrix multidomain trimeric glycoprotein is the main component of basal lamina that supports adhesion, proliferation and differentiation.

Application

Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane has been used in:
  • insulin-producing cell (IPC) differentiation.[3]
  • Infection inhibition assays.[4]
  • Proliferation Assay.[5]
Laminin supports growth and differentiation of many cell types including epithelial, endothelial, neural, muscle and liver cells. It is recommended for use as a cell culture substratum at 1-2 μg/cm2. The optimal concentration does depend on cell type as well as the application or research objectives.

Biochem/physiol Actions

Laminin proteins are integral components of structural scaffolding in animal tissues. They associate with type IV collagen via entactin and perlecan and bind to cell membranes through integrin receptors, dystroglycan glycoprotein complexes and Lutheran blood group glycoproteins. Laminin has active domains for collagen binding, cell adhesion, heparin binding, and neurite outgrowth fragment.

Components

Laminin is an extracellular matrix multidomain trimeric glycoprotein, and is the main non-collagenous component of basal lamina that supports adhesion, proliferation and differentiation. Laminin is composed of both A, B1 and B2 chains, which are connected by many disulfide bonds. This laminin product was isolated from mouse Engelbreth-Holm-Swarm tumor.

Caution

It is recommended to store this product in working aliquots at -20°C.

Preparation Note

This product is supplied at a concentration of 1 − 2 mg/mL in 50 mM Tris-HCl, with 150 mM NaCl. It is filtered using a 0.2μm filter. Thaw this solution slowly before use at 2-8°C to avoid gel formation. For use as a coating, dilute in a balanced salt solution, coat culture surface with a minimal volume and incubate at 37°C for two hours. Wash 3 times with PBS before plating cells. Laminin coatings can be stored for one month at 2-8°C.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serotype-specific entry of dengue virus into liver cells: identification of the 37-kilodalton/67-kilodalton high-affinity laminin receptor as a dengue virus serotype 1 receptor.
Thepparit C and Smith DR.
Journal of Virology, 78(22), 12647-12656 (2004)
Axonal pathfinding: Extracellular matrix role
Letourneau, P
Encyclopedia of Neuroscience, 1, 1139-1145 (2009)
Mitogenic activity of laminin on human melanoma and melanocytes: different signal requirements and role of beta 1 integrins.
Mortarini R, et al.
Cancer Research, 55(20), 4702-4710 (1995)
Alex Chia Yu Chang et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(46), 13120-13125 (2016-11-02)
Duchenne muscular dystrophy (DMD) is an incurable X-linked genetic disease that is caused by a mutation in the dystrophin gene and affects one in every 3,600 boys. We previously showed that long telomeres protect mice from the lethal cardiac disease
Julie Gaburro et al.
Emerging microbes & infections, 7(1), 68-68 (2018-04-25)
Understanding Zika virus infection dynamics is essential, as its recent emergence revealed possible devastating neuropathologies in humans, thus causing a major threat to public health worldwide. Recent research allowed breakthrough in our understanding of the virus and host pathogenesis; however

Articles

The extracellular matrix (ECM) and its attachment factor components are discussed in this article in relation to their function in structural biology and their availability for in vitro applications.

The extracellular matrix (ECM) and its attachment factor components are discussed in this article in relation to their function in structural biology and their availability for in vitro applications.

The extracellular matrix (ECM) and its attachment factor components are discussed in this article in relation to their function in structural biology and their availability for in vitro applications.

The extracellular matrix (ECM) and its attachment factor components are discussed in this article in relation to their function in structural biology and their availability for in vitro applications.

See All

Protocols

Coating surfaces with laminin for culturing cells requires specific conditions for optimal results. Protocols for coating coverslips to culture neurospheres and general cell culture are included.

Coating surfaces with laminin for culturing cells requires specific conditions for optimal results. Protocols for coating coverslips to culture neurospheres and general cell culture are included.

Coating surfaces with laminin for culturing cells requires specific conditions for optimal results. Protocols for coating coverslips to culture neurospheres and general cell culture are included.

Coating surfaces with laminin for culturing cells requires specific conditions for optimal results. Protocols for coating coverslips to culture neurospheres and general cell culture are included.

Questions

1–9 of 9 Questions  
  1. How long can stocks be frozen at -20C for?

    1 answer
    1. In undiluted form or diluted to working concentration, this material may be stored at -20°C in aliquots up to the recommended retest date for the specific lot. This data is published in the Certificate of Analysis. Avoid repeated freeze/thaw cycles. The solution may be transferred to smaller vials in working volumes for long-term storage. Plastic vials are recommended as this protein tends to adhere to glass. Please see the link below to access the product datasheet for additional details:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/409/686/l2020pis-ms.pdf

      Helpful?

  2. How will the product L2020 shipped? Will it be shipped in room temperature, on ice, or on dry ice?

    1 answer
    1. The product L2020 will be shipped using dry ice. Upon receipt, it should be stored at -20 degrees Celsius.

      Helpful?

  3. How is this laminin concentration determined? (by which method?) I tried to measure the concentration of this product by ELISA, and the results always came out half of what is written on the vial.. what is the protein purity?

    1 answer
    1. Laminin concentration is 1.0 - 2.0 mg protein/ml. The protein content is determined by Lowry method. Please refer the attached document for more details. https://www.sigmaaldrich.com/specification-sheets/131/942/L2020-1MG_________SIGMA____.pdf

      Helpful?

  4. Can I coat Laminin, Product L2020, on top of poly-L-lysine for better adherence?

    1 answer
    1. When coating coverslips or slides for use with neural stem cells (neurospheres), the following procedure is recommended. Precoat the coverslips with poly-L-lysine (Product No. P5899) at 100 μg/mL. This is incubated for 30 minutes or more at room temperature. The coverslips are then washed with sterile water and allowed to dry. The poly-lysine coated coverslips can be stored at room temperature until ready to use (up to ~1 month). Coverslips are then coated with 30 - 50 μL of warmed laminin, which is at a concentration of 50 μg/mL. Incubate at 37°C for 30 minutes. Remove the laminin, and wash with DME (50 μL) twice. Cells are then added to the coverslips.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. What type of laminin is Product L2020?

    1 answer
    1. In the nomenclature system by Burgeson, this is Laminin 1. The laminins were numbered with Arabic numerals in the order discovered. The previous A, B1, and B2 chains, and their isoforms are listed by the Greek letters, for alpha, beta, and gamma, respectively, followed by an Arabic numeral to identify the isoforms.  For example, the first laminin identified from the Engelbreth-Holm-Swarm tumor (EHS) was designated laminin-1 with the chain composition α-1/β-1/γ-1. The genes for these 3 chains are LAMA1, LAMB1, and LAMC1. Reference: Burgeson, R.E., et al., A new nomenclature for the laminins. Matrix Biol., 14, 209-211 (1994).

      Helpful?

  7. How can I store solutions of Laminin, Product L2020?

    1 answer
    1. The product has a recommended storage temperature of -20°C.  Please note, the product needs to be thawed slowly at 2-8°C.  If the product has been warmed up at room temperature and it gels, it cannot be reactivated for use. We recommend storage at 2-8°C for up to one week. For longer storage, the product needs to be frozen at -20°C in single-use aliquots.

      Helpful?

  8. What is the purity of Laminin, Product L2020?

    1 answer
    1. We do not determine a purity of this product.  An SDS PAGE gel is run and the pattern (3 bands) is comparable to past lots.

      Helpful?

  9. What is the protein concentration of the solution of Product L2020, Laminin?

    1 answer
    1. The protein specification for L2020 is 1.0 to 2.0 mg/mL. The exact value can be found on the lot-specific Certificate of Analysis, which can be downloaded from our website.

      Helpful?

Reviews

1 of 4 reviewers received a sample product or took part in a promotion
1–2 of 4 Reviews  

Active Filters

  1. Cambridge UK
    • Reviews 3
    • Votes 2
    3 out of 5 stars.

    Neural stem cell ECM

    Batch variability, supports neural stem cell attachment and maintenance.

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for your review. We encourage customers who experience problems with our products to call or email us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.

  2. Tarrytown, NY
    • Review 1
    • Vote 1
    5 out of 5 stars.

    work very well

    Both 2°C~8°C overnight coating and rapid coating(37C, 2hrs.) methods work perfectly fine

    Helpful?

  3. 2 Ratings-Only Reviews

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service