Skip to Content
Merck
All Photos(5)

Key Documents

N5751

Sigma-Aldrich

Nω-Nitro-L-arginine methyl ester hydrochloride

≥97% (TLC), powder, nitric oxide production inhibitor

Synonym(s):

L-NAME HCl, L-NAME hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15N5O4 · HCl
CAS Number:
Molecular Weight:
269.69
Beilstein:
3744166
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder

biological source

synthetic

Assay

≥97% (TLC)

form

powder

color

white to off-white

mp

132 °C

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

Cl.COC(=O)[C@@H](N)CCCNC(=N)N[N+]([O-])=O

InChI

1S/C7H15N5O4.ClH/c1-16-6(13)5(8)3-2-4-10-7(9)11-12(14)15;/h5H,2-4,8H2,1H3,(H3,9,10,11);1H/t5-;/m0./s1

InChI key

QBNXAGZYLSRPJK-JEDNCBNOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nω-Nitro-L-argininemethyl ester hydrochloride or L -NAME HCl, ananalog of arginine, inhibits NO production. It has multiple effects on thevascular system. Inhibits relaxation induced by acetylcholine and induces anincrease in arterial blood pressure. Abolishes lecithinized superoxidedismutase induced vasodilation when used to pretreat aortic ring preparationsof mice. Induces leukocyte adhesion and increases microvascular fluid andprotein fluxes and permeability. It has also been used in many studies oflearning and memory.

Application

Nω-Nitro-L-arginine methyl ester hydrochloride has been used:
  • to study the effects of brain-derived neurotrophic factor against neurotoxicity in rat cortical neurons[1]
  • to study its effect on far-infrared (FIR) enhanced microcirculation of rat skin[2]
  • to study its effect on leptin-induced regulation of myokine fibronectin type III domain containing five/irisin in murine myocytes and fat cells[3]
  • to study its effect on the levels of arginine vasopressin and neuronal nitric oxide synthase mRNA levels in hypothalamic paraventricular nucleus and supraoptic nucleus of rat[4]

Biochem/physiol Actions

Arginine analog that inhibits NO production.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Kubes et al.
The American journal of physiology, 262(2 Pt 2), H611-H615 (1992-02-01)
We recently demonstrated that inhibitors of nitric oxide (NO) production cause a dramatic increase in leukocyte adherence and emigration in postcapillary venules. The objective of this study was to assess whether inhibition of NO production leads to vascular protein leakage
L B Zou et al.
Neuropharmacology, 37(3), 323-330 (1998-07-29)
We investigated the role of nitric oxide (NO) in learning and memory formation in a radial maze test, by using the NO synthase (NOS) inhibitors, NG-nitro-L-arginine methyl ester (L-NAME) and 7-nitroindazole (7-NI), and an NO precursor, L-arginine. Rats were trained
Anne-Clémence Vion et al.
Circulation research, 112(10), 1323-1333 (2013-03-29)
Endothelial activation and apoptosis release membrane-shed microparticles (EMP) that emerge as important biological effectors. Because laminar shear stress (SS) is a major physiological regulator of endothelial survival, we tested the hypothesis that SS regulates EMP release. EMP levels were quantified
Biological effect of far-infrared therapy on increasing skin microcirculation in rats
Yu SY, et al.
Photodermatology, Photoimmunology & Photomedicine, 22(2), 78-86 (2006)
Amanda E Boe et al.
Circulation, 128(21), 2318-2324 (2013-10-05)
Long-term inhibition of nitric oxide synthase by L-arginine analogues such as N(ω)-nitro-l-arginine methyl ester (L-NAME) has been shown to induce senescence in vitro and systemic hypertension and arteriosclerosis in vivo. We previously reported that plasminogen activator inhibitor-1 (PAI-1)-deficient mice (PAI-1(-/-))

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Questions

1–5 of 5 Questions  
  1. Following reconstitution, what is the recommended storage temperature and how long it will remain stable?

    1 answer
    1. The product is tested for solubility in water at 50mg/ml. Stock solutions may be stored in aliquots at -20°C for up to 4 months.

      Helpful?

  2. What is the solution stability of Product N5751, L-NAME hydrochloride?

    1 answer
    1. Although we do not perform solution stability studies, one literature reference recommends keeping solutions on ice during the course of one day.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. How do I sterilize a prepared solution of Product N5751, L-NAME hydrochloride?

    1 answer
    1. Solutions of this product may be sterile-filtered.

      Helpful?

  5. Is Product N5751, L-NAME hydrochloride, an inhibitor of Nitric Oxide Synthase (NOS)?

    1 answer
    1. L-NAME was described as a reversible, non-specific inhibitor of nitric oxide synthase by Khavandgar. S. et al., Pharmacol. Biochem. Behav., 74, 795 (2003).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service