Skip to Content
Merck
All Photos(2)

Key Documents

N3398

Sigma-Aldrich

S-Nitroso-N-acetyl-DL-penicillamine

≥97%, powder

Synonym(s):

N-Acetyl-3-(nitrosothio)-DL-valine, S-Nitroso-N-acetylpenicillamine, SNAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12N2O4S
CAS Number:
Molecular Weight:
220.25
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97%

form

powder

color

faint green to very dark green

solubility

H2O: ≥2 mg/mL
DMSO: ≥20 mg/mL, clear (faint green to very dark green)

storage temp.

−20°C

SMILES string

CC(=O)NC(C(O)=O)C(C)(C)SN=O

InChI

1S/C7H12N2O4S/c1-4(10)8-5(6(11)12)7(2,3)14-9-13/h5H,1-3H3,(H,8,10)(H,11,12)

InChI key

ZIIQCSMRQKCOCT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

S-Nitroso-N-acetyl-DL-penicillamine (SNAP) has been used:
  • as a positive control in nitrite generation assay.
  • to block the activity of endogenous transglutaminase (TG2).
  • in proliferation assays to generate nitric oxide (NO) in cell cultures.

Biochem/physiol Actions

NO donor; vasodilator, smooth muscle relaxant, lymphocyte activator. Activates soluble guanylyl cyclase.
SNAP, a nitrosothiol derivative, releases nitric oxide (NO.) under physiological conditions thus making it a useful tool for studying pharmacological and physiological actions of NO. Because of its NO releasing properties SNAP is a potent vasodilator in vitro and in vivo and is less prone to produce pharmacological tolerance.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sophiya Garasia et al.
Scientific reports, 7(1), 1629-1629 (2017-05-11)
We assessed the association between depression status and prevalent and incident type 2 diabetes (T2D) as well as the interaction between depression and a genetic risk score (GS) based on 20 T2D single-nucleotide polymorphisms (SNPs) in a multi-ethnic longitudinal study.
Immunomodulatory activities of chitin microparticles on Leishmania major-infected murine macrophages.
Dehghani F
Archives of Medical Research, 42(7), 572-576 (2011)
Jodie D Anderson et al.
Journal of veterinary internal medicine, 33(4), 1593-1601 (2019-05-28)
Intravenous continuous rate infusion (IVCRI) of lispro at a starting dose of 0.09 U/kg/h and the use of 0.9% sodium chloride (NaCl) for fluid resuscitation in cats with diabetic ketoacidosis (DKA) have not been reported. Protocols for correction of electrolyte deficiencies
Lethal autoimmune myocarditis in interferon-gamma receptor-deficient mice: enhanced disease severity by impaired inducible nitric oxide synthase induction.
Eriksson U
Circulation, 103(1), 18-21 (2001)
The redox state of transglutaminase 2 controls arterial remodeling.
van den Akker J
PLoS ONE, 6(8), e23067-e23067 (2011)

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service