Skip to Content
Merck
All Photos(1)

Key Documents

D1797

Sigma-Aldrich

all-cis-7,10,13,16,19-Docosapentaenoic acid

synthetic, ≥97%

Synonym(s):

Clupanodonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O2
CAS Number:
Molecular Weight:
330.50
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Assay

≥97%

form

liquid

functional group

carboxylic acid

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCCCCC(O)=O

InChI

1S/C22H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h3-4,6-7,9-10,12-13,15-16H,2,5,8,11,14,17-21H2,1H3,(H,23,24)/b4-3-,7-6-,10-9-,13-12-,16-15-

InChI key

YUFFSWGQGVEMMI-JLNKQSITSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Eltweri et al.
Clinical nutrition (Edinburgh, Scotland), 36(3), 768-774 (2016-06-28)
It has been demonstrated that short term intravenous (IV) administration of omega-3 polyunsaturated fatty acids (PUFAs) is more effective than oral supplementation at promoting incorporation of the bioactive omega-3 PUFAs eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA) into plasma, blood
Jan Philipp Schuchardt et al.
Prostaglandins, leukotrienes, and essential fatty acids, 121, 76-87 (2017-06-28)
EPA and DHA cause different physiological effects, which are in many cases mediated via their oxidative metabolites (oxylipins). However, metabolism studies investigating the effect of either EPA or DHA on comprehensive oxylipin patterns are lacking. The short and long term
Additional data from our study on fatty acids variations during lactation: correlations between n-3 and n-6 PUFAs in human colostrum, transitional, and mature milk.
Carolina Moltó-Puigmartí et al.
Clinical nutrition (Edinburgh, Scotland), 30(3), 402-403 (2011-04-08)
A Voss et al.
The Journal of biological chemistry, 266(30), 19995-20000 (1991-10-25)
The hypothesis that the last step in the biosynthesis of 4,7,10,13,16,19-22:6 from linolenate is catalyzed by an acyl-CoA-dependent 4-desaturase has never been evaluated by direct experimentation. When rat liver microsomes were incubated with [1-14C]7,10,13,16,19-22:5, under conditions where linoleate was readily
Yuko Yonezawa et al.
International journal of molecular medicine, 18(4), 583-588 (2006-09-12)
We reported previously that unsaturated linear-chain fatty acids of the cis-configuration with a C18-hydrocarbon chain such as linoleic acid (cis-9, 12-octadecadienoic acid, C18:2) could potently inhibit the activity of mammalian DNA polymerases (Biochim Biophys Acta 1308: 256-262, 1996). In this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service