Skip to Content
Merck
All Photos(1)

Key Documents

44864

Supelco

cis-5,8,11,14,17-Eicosapentaenoic acid

analytical standard

Synonym(s):

EPA, Timnodonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2CH=CH)5(CH2)3CO2H
CAS Number:
Molecular Weight:
302.45
Beilstein:
1714433
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

biological source

fish oil

Quality Level

grade

analytical standard

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.4977 (lit.)

mp

−54-−53 °C (lit.)

density

0.943 g/mL at 25 °C (lit.)

format

neat

functional group

carboxylic acid

storage temp.

−20°C

SMILES string

CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCCC(O)=O

InChI

1S/C20H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h3-4,6-7,9-10,12-13,15-16H,2,5,8,11,14,17-19H2,1H3,(H,21,22)/b4-3-,7-6-,10-9-,13-12-,16-15-

InChI key

JAZBEHYOTPTENJ-JLNKQSITSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

cis-5,8,11,14,17-Eicosapentaenoic acid belongs to the class of long-chain omega-3 polyunsaturated fatty acids. It is synthesized in the human body from the essential fatty acid alpha-linolenic acid.

Application

This analyticalstandard can also be used as follows:

  • Optimization of capillary zone electrophoresis procedurecoupled with direct absorption UV detection (CZE-UV) to determine eicosapentaenoicacid and docosahexaenoic acids after their methylation in 10 different samplesof commercial omega-3 supplements
  • Simultaneous determination of eicosapentaenoic acid,docosahexaenoic acid, and arachidonic acid in human plasma samples by high-performanceliquid chromatography-electrochemical detection (HPLC-ECD)
  • Separation, identification, and quantification of eicosapentaenoicacid, docosahexaenoic acid, and arachidonic acid in human plasma samples by high-performanceliquid chromatography combined with tandem mass spectrometry (HPLC-MS/MS)

Biochem/physiol Actions

5-Lipoxygenase inhibitor; reduces thromboxane A2 production.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Calzada et al.
Biochimica et biophysica acta, 1127(2), 147-152 (1992-07-29)
The modulatory effects of eicosapentaenoic acid (EPA) on platelet arachidonic acid (AA) metabolism were applied to an in vitro model of oxidant stress. Unstimulated normal human blood platelets were first treated with a thiol-oxidizing agent, azodicarboxylic acid bis(dimethylamide) (diamide) (1
S Sipka et al.
Clinical immunology and immunopathology, 79(3), 224-228 (1996-06-01)
Free eicosapentaenoic acid (EPA) was found to inhibit dose dependently the chemiluminescence of human neutrophil granulocytes phagocytosing zymosan and their chemotaxis induced by C5a-containing zymosan-activated serum (ZAS) and platelet-activating factor. Rigidification of plasma membranes in the ZAS-treated cells could be
Richard J Woodman et al.
Atherosclerosis, 166(1), 85-93 (2002-12-17)
Type 2 diabetes and hypertension are both associated with an increased risk of atherothrombosis. We assessed whether purified eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA) from fish oil have differential effects on platelet, fibrinolytic and vascular function in patients with
Cindy A Francois et al.
The American journal of clinical nutrition, 77(1), 226-233 (2002-12-25)
Flaxseed oil is a rich source of 18:3n-3 (alpha-linolenic acid, or ALA), which is ultimately converted to 22:6n-3 (docosahexaenoic acid, or DHA), a fatty acid important for the development of the infant brain and retina. The objective of this study
Wendy L Hall et al.
The Journal of nutrition, 138(2), 287-291 (2008-01-22)
Diets rich in eicosapentaenoic acid [EPA; 20:5(n-3)] are associated with decreased arterial stiffness, but postprandial effects on vascular function are unknown. We investigated whether an EPA-enriched high-fat meal could improve postprandial vascular function. Seventeen healthy men ingested 2 test meals

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service