Skip to Content
Merck

E4504

cis-8,11,14-Eicosatrienoic acid

≥99%

Synonym(s):

bis-Homo-γ-linolenic acid, Homo-γ-linolenic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)3(CH2CH=CH)3(CH2)6CO2H
CAS Number:
Molecular Weight:
306.48
UNSPSC Code:
12352211
NACRES:
NA.25
PubChem Substance ID:
EC Number:
217-233-0
Beilstein/REAXYS Number:
1796533
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

plant oil (Borage)

Quality Segment

assay

≥99%

form

viscous liquid

refractive index

n20/D 1.478 (lit.)

functional group

carboxylic acid

lipid type

omega FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/CCCCCCC(O)=O

InChI

1S/C20H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10,12-13H,2-5,8,11,14-19H2,1H3,(H,21,22)/b7-6-,10-9-,13-12-

InChI key

HOBAELRKJCKHQD-QNEBEIHSSA-N



Still not finding the right product?

Explore all of our products under cis-8,11,14-Eicosatrienoic acid


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

143.6 °F - closed cup

flash_point_c

62 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library