Skip to Content
Merck
All Photos(4)

Key Documents

C7527

Sigma-Aldrich

Choline chloride

BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%

Synonym(s):

(2-Hydroxyethyl)trimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(Cl)CH2CH2OH
CAS Number:
Molecular Weight:
139.62
Beilstein:
3563126
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.75
Pricing and availability is not currently available.

biological source

synthetic (organic)

product line

BioReagent

Assay

≥98%

form

crystals

mol wt

Mw 139.62 g/mol

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable
cell culture | plant: suitable

color

white

pH

5.0-6.5 (25 °C, 140 g/L)

mp

302-305 °C (dec.) (lit.)

solubility

H2O: 50 mg/mL

SMILES string

[Cl-].C[N+](C)(C)CCO

InChI

1S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1

InChI key

SGMZJAMFUVOLNK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Choline chloride is a quaternary ammonium salt and associates with hydrogen bond donors (HBD).[1] It is essential for the vital physiological functions and is used as a nutritional additive. Choline is a source of methyl groups for methyl-transfer reactions.[2]

Application

Choline chloride has been used:
  • as a component of choline acetyltransferase (ChAT) buffer and for choline uptake in cultured human neuroblastoma (SK-N-SH) cells[3]
  • for choline standard curve generation for quantifying phospholipase D (PLD) activity[4]
  • as a component of slice solution for the dissection of entorhinal-hippocampal brain slices[5]

Substrates

Choline acetyltransferase substrate

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of choline chloride based binary mixtures
Abbas Q and Binder L
ECS Transactions, 33(7), 49-59 (2010)
Apoptosis of bovine granulosa cells: Intracellular pathways and differentiation
Carou MC, et al.
Acta Histochemica, 119(5), 462-470 (2017)
Harvie P Portugaliza et al.
Scientific reports, 9(1), 14595-14595 (2019-10-12)
Transmission of malaria parasites from humans to mosquito vectors requires that some asexual parasites differentiate into sexual forms termed gametocytes. The balance between proliferation in the same host and conversion into transmission forms can be altered by the conditions of
Choline: an essential nutrient for public health
Zeisel SH and Da Costa KA
Nutr. Rev, 67(11), 615-623 (2009)
Functional calcium imaging in developing cortical networks
Dawitz J, et al.
Journal of Visualized Experiments, 119(56), e3550-e3550 (2011)

Questions

1–3 of 3 Questions  
  1. Choline chloride will be melted in water for addition to the medium. Can you please tell me the proper storage method, such as -30°C freezing ay -4°C refrigeration?

    1 answer
    1. This product is soluble in water at 50 mg/mL. The storage and stability conditions for this product upon reconstitution have not been determined.

      Helpful?

  2. Hi, is it possible to thaw choline chloride in PBS instead of water and add it to the medium?

    1 answer
    1. This product is soluble in water, tested at 50 mg/mL. Solubility in PBS has not been determined in-house. Literature suggests that choline chloride can be prepared with PBS. For reference, see https://en-cdn.bio-protocol.org/pdf/Bio-protocol807.pdf?rel_link=YmlvLXByb3RvY29sLm9yZy9lbi9icGRldGFpbD9pZD04MDcmdHlwZT0w

      Helpful?

  3. Hi, could you please provide the product information sheet for Choline chloride.

    1 answer
    1. There is no specific product information sheet offered for Choline Chloride. Please see the PROPERTIES section of the product page for details. Links to a sample Certificate of Analysis and a Certificate of Origin are provided below:

      Certificate of Analysis
      https://www.sigmaaldrich.com/certificates/COFA/C7/C7527/C7527-100G________SLBB8921V_.pdf

      Certificate of Origin
      https://www.sigmaaldrich.com/US/en/coo/undefined/C7527/SLBB8921V

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service