Skip to Content
Merck
All Photos(3)

Key Documents

C3400

Sigma-Aldrich

Casein from bovine milk

powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

bovine milk

Assay

87-94% protein basis

form

powder

technique(s)

activity assay: suitable
electrophoresis: suitable
immunocytochemistry: suitable

mp

280 °C (dec.) (lit.)

solubility

H2O: insoluble (forms a cloudy suspension)

UniProt accession no.

SMILES string

[P](=O)(OCC(NC(=O)C(NC(=O)C(N)Cc1ccccc1)CCC(=O)N)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(CCC(=O)N)C(=O)NC(C(O)C)C(=O)NC(CCC(=O)O)C(=O)NC(CC(=O)O)C(=O)NC(CCC(=O)O)C(=O)NC(CC(C)C)C(=O)NC(CCC(=O)N)C(=O)NC(CC(=O)O)C(=O)NC(C

InChI

1S/C81H125N22O39P/c1-36(2)31-50(76(132)94-43(15-24-57(87)108)71(127)101-52(34-64(120)121)78(134)98-49(81(137)138)11-7-8-30-82)99-72(128)47(19-28-61(114)115)95-77(133)51(33-63(118)119)100-73(129)48(20-29-62(116)117)97-80(136)65(37(3)104)103-75(131)44(16-25-58(88)109)92-68(124)42(14-23-56(86)107)90-67(123)41(13-22-55(85)106)91-69(125)45(17-26-59(110)111)93-70(126)46(18-27-60(112)113)96-79(135)53(35-142-143(139,140)141)102-74(130)40(12-21-54(84)105)89-66(122)39(83)32-38-9-5-4-6-10-38/h4-6,9-10,36-37,39-53,65,104H,7-8,11-35,82-83H2,1-3H3,(H2,84,105)(H2,85,106)(H2,86,107)(H2,87,108)(H2,88,109)(H,89,122)(H,90,123)(H,91,125)(H,92,124)(H,93,126)(H,94,132)(H,95,133)(H,96,135)(H,97,136)(H,98,134)(H,99,128)(H,100,129)(H,101,127)(H,102,130)(H,103,131)(H,110,111)(H,112,113)(H,114,115)(H,116,117)(H,118,119)(H,120,121)(H,137,138)(H2,139,140,141)

InChI key

BECPQYXYKAMYBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Casein from bovine milk is a phosphoprotein and forms three-dimensional colloidal supramolecular micelles. There are four main types of casein which make up approximately 80% of the total protein in bovine milk: α-s1 casein, α-s2 casein, β-casein, and κ-casein. Casein is proposed to be the main protective constituent in milk. Casein is an amphiphilic protein.

Application

Casein from bovine milk has been used:
  • as a solid food sample in the in vivo and the in vitro digestion experiments using rodents
  • to prepare P407-casein hydrogels and to study the mechanical effects of the addition of casein to P407
  • in the preincubation solution, to increase the photostability of the quantum dots and to decrease nonspecific binding, for real-time imaging of single synaptic vesicles in hippocampal neurons

Casein from bovine milk is a phosphoprotein. There are four main types of Casein which make up approximately 80% of the total protein in bovine milk: α-s1 Casein, α-s2 Casein, β-Casein, and κ-Casein. Casein is proposed to be the main protective constituent in milk.

Biochem/physiol Actions

Casein is useful in food industries and non-food applications. It has the property for emulsification, foam formation, and stabilization, water-binding, and gelation. Casein is also considered heat and acid stable. It can serve as an indispensable diet for rodents.
Partial gastrointestinal digestion of casein is a rich source of bioactive peptides, such as β-casomorphin. However, bovine casein is not homogeneous; variants A1 and B do lead to production of β-casomorphin 7 production, while A2 does not.

Quality

Essentially vitamin free.

Preparation Note

Lactic acid precipitated New Zealand casein extracted with ethyl alcohol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Encyclopedia of Food Chemistry (2019)
Zealyn Shi-Lin Heng et al.
Antibody therapeutics, 5(1), 30-41 (2022-02-12)
Optimizing recombinant antibody production is important for cost-effective therapeutics and diagnostics. With impact on commercialization, higher productivity beyond laboratory scales is highly sought, where efficient production can also accelerate antibody characterizations and investigations. Investigating HEK293E cells for mammalian antibody production
Kashish Jain et al.
Nature protocols, 17(10), 2275-2306 (2022-07-28)
The spatiotemporal aspects of early signaling events during interactions between cells and their environment dictate multiple downstream outcomes. While advances in nanopatterning techniques have allowed the isolation of these signaling events, a major limitation of conventional nanopatterning methods is its
Tauqeerunnisa Syeda et al.
Journal of Alzheimer's disease : JAD, 66(4), 1657-1682 (2018-11-27)
Recent investigations have demonstrated an important role of gut microbiota (GM) in the pathogenesis of Alzheimer's disease (AD). GM modulates a host's health and disease by production of several substances, including lipopolysaccharides (LPS) and short-chain fatty acids (SCFAs), among others.
Impact of dietary proteins on energy balance, insulin sensitivity, and glucose homeostasis: From proteins to peptides to amino acids
The Molecular Nutrition of Amino Acids and Proteins, 241-264 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service