Skip to Content
Merck
All Photos(3)

Key Documents

P5155

Sigma-Aldrich

D-Pantothenic acid hemicalcium salt

BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture

Synonym(s):

(R)-(+)-N-(2,4-Dihydroxy-3,3-dimethyl-1-oxobutyl)-β-alanine hemicalcium salt, Calcium D-pantothenate, Vitamin B5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2C(CH3)2CH(OH)CONHCH2CH2CO2 ·1/2Ca
CAS Number:
Molecular Weight:
238.27
Beilstein:
3769272
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058012
PubChem Substance ID:
NACRES:
NA.75
Pricing and availability is not currently available.

biological source

synthetic (organic)

product line

BioReagent

Assay

≥98% (TLC)

form

powder

mol wt

Mw 238.27 g/mol

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable
cell culture | plant: suitable

color

white to off-white

pH

7.0-8 (25 °C, 50 g/L)

solubility

water: soluble 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)O[Ca]OC(=O)CCNC(=O)[C@H](O)C(C)(C)CO

InChI

1S/2C9H17NO5.Ca/c2*1-9(2,5-11)7(14)8(15)10-4-3-6(12)13;/h2*7,11,14H,3-5H2,1-2H3,(H,10,15)(H,12,13);/q;;+2/p-2/t2*7-;/m00./s1

InChI key

FAPWYRCQGJNNSJ-UBKPKTQASA-L

Looking for similar products? Visit Product Comparison Guide

General description

Plants and microorganisms, such as Escherichia coli are the major sources of D-pantothenic acid.[1] It is water soluble[2] and quite stable to heating.[3]

Application

D-Pantothenic acid hemicalcium salt has been used:
  • as a component of FMX-turbodoma- derived media[2]
  • as a supplement in DMEM (Dulbecco′s modified eagle media)/F-12 used for cell proliferation[4]
  • as a supplement in preadipocyte media[5]

Precursor in the biosynthesis of coenzyme A.

Biochem/physiol Actions

Along with coenzyme A (CoA, D-pantothenic acid is a precursor for acyl carrier protein.[1] These are essential cofactors[1] involved in fat, protein and carbohydrate metabolism. It also participates in the formation of acetylcholine neurotransmitter. D-pantothenic acid is considered as a source of nutrition and possesses immune function by mediating antibody formation. The production of cortisone is supported by D-pantothenic acid. It is essential for adrenal gland activity.[3]

Physical properties

Due to the unstable, hygroscopic nature of the free acid, the calcium salt is employed.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3T3-L1 cells and perivascular adipocytes are not equivalent in amine transporter expression
Ismail A, et al.
Febs Letters, 591(1), 137-144 (2017)
Stopping the Clock, 184-184 (2003)
The role of vitamins and amino acids on hybridoma growth and monoclonal antibody production
Ducommun P, et al.
Cytotechnology, 37(2), 65-73 (2001)
LipiD-QuanT: a novel method to quantify lipid accumulation in live cells.
Varinli H, et al.
Journal of Lipid Research, 56(11), 2206-2216 (2015)
Organisation of the pantothenate (vitamin B5) biosynthesis pathway in higher plants
Ottenhof HH, et al.
The Plant Journal, 37(1), 61-72 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service