Skip to Content
Merck
All Photos(2)

Key Documents

B8667

Sigma-Aldrich

Bovine Serum Albumin solution

20 mg/mL in H2O, low bioburden, protease free, for molecular biology

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.27

biological source

bovine

Quality Level

grade

Molecular Biology
for molecular biology

product line

BioReagent

Assay

20-22 mg/mL protein basis (biuret)

form

solution

quality

non-acetylated

purified by

heat shock fractionation

contains

does not contain preservative

origin

USA origin

concentration

20 mg/mL in H2O
20-22 mg/mL Biuret

technique(s)

PCR: suitable

impurities

Alakaline phosphatase, none detected
DNase, none detected
Endonuclease, none detected
Exonuclease, none detected
NICKase, none detected
peroxidase, none detected
<10.0 cfu/mL Total microbial count

UniProt accession no.

foreign activity

Alkaline Phosphatase free
DNase free
DNase, RNase, NICKase, protease, peroxidase and alkaline phosphatase, none detected
NICKase free
Peroxidase free
Protease free
RNase free

shipped in

dry ice

storage temp.

−20°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

Application

Select the right albumin for your application with the Albumin Selection Guide.
BSA has various uses in molecular biology applications, such as:
  • As an added component in solutions of enzymes or of other proteins, to prevent adhesion of the enzyme or other protein of interest to the reaction tube surface or to pipette surfaces
  • As an inert enzyme or protein stabilizer during incubation, to allow the enzyme or protein of interest to function properly

Features and Benefits

This BSA solution product for molecular biology is tested for the following properties:
  • Alkaline Phosphatase-free
  • DNase-free / Exonuclease-free
  • Nickase-free / Endonuclease-free
  • Protease-free
  • RNase-free

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Beam et al.
Clinical transplantation, 32(1) (2017-11-08)
Cytomegalovirus (CMV) pneumonia causes major morbidity and mortality. Its diagnosis requires demonstration of viral cytopathic changes in tissue, entailing risks of lung biopsy. This study aimed to determine CMV viral load (VL) thresholds in bronchoalveolar lavage fluid (BALF) for diagnosis
Alexander S Day et al.
Scientific reports, 11(1), 19933-19933 (2021-10-09)
An emulsion loop-mediated isothermal amplification (eLAMP) platform was developed to reduce the impact that contamination has on assay performance. Ongoing LAMP reactions within the emulsion droplets cause a decrease in interfacial tension, causing a decrease in droplet size, which results
Anjelika Gasilina et al.
Bio-protocol, 12(18) (2022-11-01)
Understanding the molecular and structural mechanisms that govern the assembly and organization of higher-order actin architecture requires the use of in vitro actin binding and bundling assays. Crosslinking of actin filaments into bundles can be monitored in vitro via several
Martin Frappier et al.
Probiotics and antimicrobial proteins, 14(5), 873-883 (2022-06-16)
Dyslipidemia, specifically abnormal levels of low-density lipoprotein cholesterol (LDL-C), is an important risk factor of cardiovascular disease. Evidence showing the promising abilities of probiotics to lower total cholesterol or LDL-C has, however, not yet convinced experts to recommend probiotic bacteria
Naveen Ramalingam et al.
Biomicrofluidics, 11(3), 034110-034110 (2017-06-15)
Bovine serum albumin (BSA) is widely used as an additive in polymerase chain reaction (PCR)-based microfluidic devices to passivate reactors and alleviate nucleic-acid amplification. BSA is available commercially in two types: either acetylated or non-acetylated. A survey of literature indicates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service