Skip to Content
Merck
All Photos(1)

Key Documents

83266

Sigma-Aldrich

Magnesium sulfate solution

BioUltra, for molecular biology

Synonym(s):

Magnesium Sulphate, Magnesium(II) sulfate, MgSO4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgSO4
CAS Number:
Molecular Weight:
120.37
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
grade:
for molecular biology
form:
liquid

grade

for molecular biology

Quality Level

product line

BioUltra

form

liquid

concentration

2.5 M in H2O

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
<0.002% total nitrogen (N)

pH

6.5-8.5 (25 °C)

anion traces

chloride (Cl-): ≤5 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤20 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.03
λ: 280 nm Amax: 0.02

SMILES string

[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

CSNNHWWHGAXBCP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Optimization in vinasse processing: Magnesium sulfate plays a role in optimizing the electrodialysis and nanofiltration process of vinasse, enhancing environmental sustainability and efficiency in waste management practices, which are crucial for industrial applications including the production of IVD reagents (Duarte et al., 2023).

  • Screening of flame retardant residues: In analytical methodologies like QuEChERS-gas chromatography, magnesium sulfate is employed for its efficacy in the extraction and cleanup steps, underscoring its importance in ensuring high-purity standards required for accurate and reliable diagnostic assays (Li et al., 2023).

Biochem/physiol Actions

Magnesium sulphate can be used to treat acute severe asthma. It is mainly considered as a neuroprotective agent.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cardiovascular Disease
Handbook on the Toxicology of Metals, 801, 313-331 (2015)
Effects of nebulised magnesium sulphate on inflammation and function of the guinea-pig airway
Turner DL, et al.
European Journal of Pharmacology, 801, 79-85 (2017)
Lex W Doyle
Current opinion in pediatrics, 24(2), 154-159 (2012-01-10)
Antenatal magnesium sulfate may reduce the excessive rates of cerebral palsy in survivors of very preterm birth. There are five randomized controlled trials of magnesium sulfate therapy given to the mother prior to very preterm birth which have reported neurological
Robert J Floor et al.
Chembiochem : a European journal of chemical biology, 15(11), 1660-1672 (2014-07-01)
We explored the use of a computational design framework for the stabilization of the haloalkane dehalogenase LinB. Energy calculations, disulfide bond design, molecular dynamics simulations, and rational inspection of mutant structures predicted many stabilizing mutations. Screening of these in small
Claire L O'Brien et al.
Gut, 63(10), 1596-1606 (2014-01-17)
Our aim was to determine whether or not specific microorganisms were transported selectively to lymph nodes in Crohn's disease (CD) by comparing node and mucosal microbial communities in patients and controls. We also sought evidence of dysbiosis and bacterial translocation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service