Skip to Content
Merck
All Photos(1)

Key Documents

10711454001

Roche

Bovine Serum Albumin

blood typing: suitable, Endonuclease, Exonuclease, Rnases, Proteinases free

Synonym(s):

Bovine Serum Albumin, BSA, Bovine albumin, Bovine serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

bovine

Quality Level

form

solution

packaging

pkg of 1 mL (20 mg/ml)

manufacturer/tradename

Roche

technique(s)

DNA synthesis: suitable

pH

4.8-7.5

foreign activity

RNase, none detected
endonuclease, none detected
exonuclease, none detected
proteinase, none detected

storage temp.

−20°C (−15°C to −25°C)

Looking for similar products? Visit Product Comparison Guide

General description

The product, bovine serum albumin, is of special quality for molecular biology. Bovine serum albumin has homology with human serum albumin. Serum albumins are soluble proteins of the circulatory system and participate in various cellular processes.

Contents
Solution of 20mg/ml Bovine Serum Albumin in 50mM Tris-HCl, 0.1M NaCl, 0.25mM EDTA, 1mM 2-mercaptoethanol, 50% glycerol (v/v), pH approximately 7.6.

Application

Bovine Serum Albumin supports enzyme stability. It has been used for the:
  • preparation of circular DNA molecules
  • amplification of bacterial 16S rRNA genes
  • BOX-PCR fingerprinting

Quality

Contaminants: Albumin is checked for the absence of DNases, nicking activity, RNases, and proteases according to the current Quality Control procedures.

Physical form

Solution of 20 mg/ml Bovine Serum Albumin in 50 mM Tris-HCl, 0.1 M NaCl, 0.25 mM EDTA, 1 mM 2-mercaptoethanol, 50% glycerol (v/v), pH approx. 7.6.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soledad Sagastume et al.
The Journal of eukaryotic microbiology, 61(1), 42-50 (2013-10-10)
To date, few organisms have been shown to possess variable ribosomal RNA, otherwise considered a classic example of uniformity by concerted evolution. The polymorphism for the 16S rRNA in Nosema ceranae analysed here is striking as Microsporidia are intracellular parasites
P Bourassa et al.
The journal of physical chemistry. B, 114(9), 3348-3354 (2010-02-13)
We report the complexation of bovine serum albumin (BSA) with resveratrol, genistein, and curcumin, at physiological conditions, using constant protein concentration and various polyphenol contents. FTIR, CD, and fluorescence spectroscopic methods were used to analyze the ligand binding mode, the
Kristopher D Barbee et al.
Macromolecular bioscience, 11(5), 607-617 (2011-02-10)
A method for fabricating DNA polymer brush arrays using photolithography and plasma etching followed by solid-phase enzymatic DNA amplification is reported. After attaching oligonucleotide primers to the surface of a glass coverslip, a thin layer of photoresist is spin-coated on
Guido Alberto Gnecchi-Ruscone et al.
Cell, 185(8), 1402-1413 (2022-04-03)
The Avars settled the Carpathian Basin in 567/68 CE, establishing an empire lasting over 200 years. Who they were and where they came from is highly debated. Contemporaries have disagreed about whether they were, as they claimed, the direct successors
Xavier Fernandez-Cassi et al.
Water research, 200, 117252-117252 (2021-05-29)
Wastewater-based epidemiology (WBE) has been shown to coincide with, or anticipate, confirmed COVID-19 case numbers. During periods with high test positivity rates, however, case numbers may be underreported, whereas wastewater does not suffer from this limitation. Here we investigated how

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service