Skip to Content
Merck
All Photos(1)

Key Documents

A7979

Sigma-Aldrich

Bovine Serum Albumin solution

35% in DPBS, sterile-filtered, BioXtra, suitable for cell culture

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

biological source

bovine

Quality Level

description

endotoxin tested

sterility

sterile-filtered

product line

BioXtra

Assay

33-37% total protein basis (UV)

form

liquid

origin

USA origin

concentration

35% in DPBS

technique(s)

blocking: suitable
cell culture | mammalian: suitable

impurities

≤1000 EU/mL Endotoxin level

pH

6.5-7.5

foreign activity

Mycoplasma Test by Barile On raw material, none detected

shipped in

ambient

storage temp.

2-8°C

General description

BSA is a single polypeptide chain consisting of about 583 amino acid residues and no carbohydrates. At pH 5-7 it contains 17 intrachain disulfide bridges and 1 sulfhydryl group.

Application

Bovine serum albumin is broadly used as an additive to cell culture media, especially serum-free media. It provides a range of benefits including protection from oxidative damage and stabilization of other media components such as fatty acids and pyridoxal.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina Janssen et al.
Methods in molecular biology (Clifton, N.J.), 1994, 31-39 (2019-05-28)
To better understand and model neurological, in particular neurodegenerative diseases, human induced pluripotent stem cells (hiPSCs) offer a great source for generation of neural cells. We provide a protocol for the differentiation of hiPSc-derived astrocytes in vitro. This protocol not
Brett S Peterson et al.
Cell reports, 24(1), 209-223 (2018-07-05)
SIRT3 is a nicotinamide adenine dinucleotide (NAD+)-dependent mitochondrial protein deacetylase purported to influence metabolism through post-translational modification of metabolic enzymes. Fuel-stimulated insulin secretion, which involves mitochondrial metabolism, could be susceptible to SIRT3-mediated effects. We used CRISPR/Cas9 technology to manipulate SIRT3 expression in
Mohammed Samer Shaban et al.
Nature communications, 12(1), 5536-5536 (2021-09-22)
Coronaviruses (CoVs) are important human pathogens for which no specific treatment is available. Here, we provide evidence that pharmacological reprogramming of ER stress pathways can be exploited to suppress CoV replication. The ER stress inducer thapsigargin efficiently inhibits coronavirus (HCoV-229E
Gustavo A Gomez et al.
Developmental biology, 449(2), 99-106 (2019-03-04)
The developmental biology of neural crest cells in humans remains unexplored due to technical and ethical challenges. The use of pluripotent stem cells to model human neural crest development has gained momentum. We recently introduced a rapid chemically defined approach
John P Ray et al.
Nature communications, 11(1), 1237-1237 (2020-03-08)
Genome-wide association studies have associated thousands of genetic variants with complex traits and diseases, but pinpointing the causal variant(s) among those in tight linkage disequilibrium with each associated variant remains a major challenge. Here, we use seven experimental assays to

Articles

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Explore serum albumin's role in serum-free cell culture systems for biomanufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service