Skip to Content
Merck
All Photos(1)

Key Documents

A9544

Sigma-Aldrich

Anti-Human IgG (Fc specific)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti Human Igg, Anti-Human IgG Antibody - Anti-Human IgG (Fc specific)-Alkaline Phosphatase antibody produced in goat, Anti-Human Igg, anti human IgG antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:55,000
dot blot: 1:40,000 (chemiluminescent)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:150

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Related Categories

General description

Human IgGs are glycoprotein antibodies that contain two equivalent light chains and a pair of identical heavy chains. IgGs have four distinct isoforms, ranging from IgG1 to IgG4. Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. IgG is usually found as a monomer. IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. About 70 percent of the total immunoglobulin consists of IgG. Immunoglobulin G (IgG) participates in hypersensitivity type II and type III. Anti-Human IgG (Fc specific)-Alkaline Phosphatase antibody is specific for human IgG and human IgG, Fc fragment. The product does not react with human IgG, Fab fragment, light chains, IgA, and IgM. Additionally, the antibody does not bind to mouse and rat IgG. Goat anti-human IgG is conjugated to alkaline phosphatase by protein cross linking with 0.2% glutaraldehyde.

Specificity

Specificity of the Anti-Human IgG is determined prior to conjugation by ELISA. The conjugate is specific for human IgG and human IgG, Fc fragment. Cross reactivity of the antibody-conjugate is also determined by ELISA. The conjugate shows no reactivity with human IgG, Fab fragment, human light chains, human IgA, human IgM or with mouse IgG and rat IgG.
Binds human IgG; does not bind other human Igs.
Provides reduced background with mouse or rat samples.

Immunogen

Human IgG, Fc fragment

Application

A modified monoclonal antibody specific immobilization of platelet antigens (MAIPA) assay was performed on human platelet eluates using alkaline phosphatase-conjugated goat anti-human, Fc specific, antibody as the secondary with a 30 minute incubation.
Anti-Human IgG (Fc specific)-Alkaline Phosphatase antibody is suitable for use in ELISA and western blot at 1:5000 dilutions. The product can also be used for dot blot (1:40,000) and immunohistochemistry (1:150 using formalin-fixed, paraffin-embedded sections).

Anti-Human IgG (Fc specific)-Alkaline Phosphatase antibody has been used in
  • quantitative sandwich ELISA
  • α-GalC ELISA
  • ELISA assays
  • ELISAs for IgG subclasses

Biochem/physiol Actions

Immunoglobulin G (IgG) regulates immunological responses to allergy and pathogenic infections. IgGs have also been implicated in complement fixation and autoimmune disorders. Immunoglobulin G (IgG) mainly participates in hypersensitivity type II and type III.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% BSA, 1 mM MgCl2 and 15 mM sodium azide

Storage and Stability

Store at 2-8 °C. Do Not Freeze.

Other Notes

No cross-reaction with mouse and rat IgG.
Working dilutions should be determined by titration assay. Due to differences in assay systems, these titers may not reflect the user′s actual working dilution.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Puntoriero et al.
The EMBO journal, 17(13), 3521-3533 (1998-07-03)
The hypervariable region 1 (HVR1) of the putative envelope protein E2 of hepatitis C virus (HCV) is the most variable antigenic fragment in the whole viral genome and is mainly responsible for the large inter-and intra-individual heterogeneity of the infecting
Ming Hou et al.
European journal of haematology, 70(6), 353-357 (2003-05-21)
The treatment of chronic idiopathic thrombocytopenic purpura (ITP) is difficult in those unresponsive to corticosteroids and/or splenectomy. We attempted to induce durable response in 21 patients with refractory ITP by applying mycophenolate mofetil (MMF) (1.5-2.0 g/d), a novel immunosuppressive agent.
A Juhász et al.
Journal of medical virology, 64(4), 526-530 (2001-07-27)
Sera of blood donors were investigated by a peptide ELISA and indirect immunofluorescence assay to assess the prevalence of HHV-8 infection in the Hungarian population. A 14 amino acid long synthetic oligopeptide from the carboxyterminus of orf65/small virus capsid antigen
Wanwisa Dejnirattisai et al.
Nature immunology, 17(9), 1102-1108 (2016-06-25)
Zika virus (ZIKV) was discovered in 1947 and was thought to lead to relatively mild disease. The recent explosive outbreak of ZIKV in South America has led to widespread concern, with reports of neurological sequelae ranging from Guillain Barré syndrome
M F Saeed et al.
Journal of clinical microbiology, 39(7), 2445-2452 (2001-06-28)
Oropouche (ORO) virus is an emerging infectious agent that has caused numerous outbreaks of an acute febrile (dengue-like) illness among humans in Brazil, Peru, and Panama. Diagnosis of ORO virus infection is based mainly on serology. Two different antigens, hamster

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service