Skip to Content
Merck
All Photos(1)

Key Documents

A3312

Sigma-Aldrich

Anti-Human IgG (γ-chain specific), F(ab′)2 fragment−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

direct ELISA: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50
western blot: 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Human IgGs are glycoprotein antibodies that contain two equivalent light chains and a pair of identical heavy chains. IgGs have four distinct isoforms, ranging from IgG1 to IgG4. These antibodies regulate immunological responses to allergy and pathogenic infections. IgGs have also been implicated in complement fixation and autoimmune disorders . Anti-Human IgG (γ-chain specific), (F(ab′)2) fragment-Alkaline Phosphatase antibody is specific for human IgG when tested against human IgA, IgG, IgM, and Bence Jones κ and λ myeloma proteins.

Immunogen

Purified human IgG

Application

Goat anti-human IgG (gamma-chain specific), F(ab′)2 fragment-alkaline phosphatase antibody can be used for western blot (1:30,000) applications.
Anti-Human IgG (γ-chain specific), (F(ab′)2) fragment-Alkaline Phosphatase antibody is suitable for use in ELISA and immunohistochemistry.
Serum IgGs against HHV-6 or HHV-7 were detected by Elisa using alkaline phosphatase conjugated goat anti-human IgG F′ab specific at a concentration of 1:1000 diluted in PBS/0.05% skim milk. The antibody was incubated on plates for 2 hours at 37 degrees.

Physical form

Solution in 0.05 M Tris buffer, pH 8.0, containing 1 mM MgCl2, 10 mM glycine, 1% bovine serum albumin, 50% glycerol and 15 mM sodium azide as preservative

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer L Chain et al.
Frontiers in psychiatry, 11, 564-564 (2020-07-17)
Movement, behavioral, and neuropsychiatric disorders in children have been linked to infections and a group of anti-neuronal autoantibodies, implying dopamine receptor-mediated encephalitis within the basal ganglia. The purpose of this study was to determine if anti-neuronal biomarkers, when used as
T J Sims et al.
Oral microbiology and immunology, 14(2), 73-85 (1999-04-29)
We obtained clinical isolates of Porphyromonas gingivalis of known ribotype from patients diagnosed with adult periodontitis and used Western blot methodology to evaluate profiles of antigens recognized by IgG in heterologous and homologous patient sera. Our aims were to identify
Kathleen Connery et al.
Translational psychiatry, 8(1), 148-148 (2018-08-12)
The identification of brain-targeted autoantibodies in children with autism spectrum disorder (ASD) raises the possibility of autoimmune encephalopathy (AIE). Intravenous immunoglobulin (IVIG) is effective for AIE and for some children with ASD. Here, we present the largest case series of
Sabrina Matà et al.
Journal of the peripheral nervous system : JPNS, 9(3), 138-143 (2004-09-15)
Few reports exist on the association between the humoral immune response to glycolipids and neuropathic findings in diabetes. To address this issue, we assayed serum anti-GM1, GD1b, GD1a, and sulfatides IgG and IgM in a group of 85 non-selected diabetic
T J Sims et al.
Journal of clinical immunology, 18(5), 355-367 (1998-10-30)
Bacteroides forsythus is one of the etiologic agents of destructive periodontal diseases. Determining which antigenic components of the bacterium are recognized in the immune response of periodontitis patients is an important step in assessing strategies for vaccine development. The aim

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service