Skip to Content
Merck
All Photos(1)

Key Documents

A0705

Sigma-Aldrich

Anti-Bovine IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Rabbit Anti-Bovine IgG (whole molecule)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

bovine

technique(s)

direct ELISA: 1:30,000
dot blot: 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulins (Igs) belongs to immunoglobulin super-family (IgSF). Igs in general, consist of two heavy (H) chains and two light (L) chains. The L chain can be either κ or a λ chain. Each chain contains one NH2-terminal “variable" (V) region and one or more COOH-terminal “constant” (C) regions. Each of which consists of two sandwiched β pleated sheets ′pinned′ together by a disulfide bridge. There are five main classes of Igs based on heavy chain C domains, they are, IgM, IgG, IgA, IgD, and IgE isotypes. IgG can be divided into four subclasses, IgG1, IgG2, IgG3, and IgG4. Each subclass possesses distinct biologic properties.

Immunogen

Purified bovine IgG

Application

Anti-Bovine IgG (whole molecule)-Alkaline Phosphatase antibody produced in rabbit has been used in
  • enzyme linked immunosorbent assay (ELISA)
  • development of immunosensors
  • western blot analysis

Biochem/physiol Actions

Bovine IgGs are glycoprotein antibodies that regulate immune responses in herds. These antibodies inhibit TLR5 activation upon immunization with native H7 flagellin . Bovine IgG levels can be used for the detection of Johne′s disease and milk adulteration .

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2, 50% glycerol, and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of lactoferrin and immunoglobulin G in animal milks by new immunosensors
Campanella L, et al.
Sensors, 9(3), 2202-2221 (2009)
Detection of Bovine IgG Isotypes in a PPA-ELISA for Johne's Disease Diagnosis in Infected Herds
Fernandez, B., et al.
Veterinary Medicine International, doi: 10-doi: 10 (2012)
Sarfraz-Ur-Rahman et al.
Animals : an open access journal from MDPI, 12(16) (2022-08-27)
Toxoplasma gondii is a protozoan parasite that causes toxoplasmosis in warm-blooded vertebrates, globally. The main aims of this study were to assess the seropositivity to toxoplasmosis of an exotic breed of cattle (n = 400) from different farms using the
José Gomes Pereira et al.
Revista brasileira de parasitologia veterinaria = Brazilian journal of veterinary parasitology : Orgao Oficial do Colegio Brasileiro de Parasitologia Veterinaria, 30(4), e014321-e014321 (2021-12-16)
Anaplasma marginale is an obligate intracellular Gram-negative bacterium found in ruminants' erythrocytes and is the etiological agent of bovine anaplasmosis. The bacterium's genetic diversity has been characterized based on sequences of major surface proteins (MSPs), such as MSP1α. The aim
Identification and analysis of immunodominant antigens for ELISA-based detection of Theileria annulata
Bilgic HB, et al.
Testing, 11(6), e0156645-e0156645 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service