Skip to Content
Merck
All Photos(2)

Key Documents

B1911

Sigma-Aldrich

BCIP®/NBT

alkaline phosphatase substrate, chromogenic, liquid

Synonym(s):

5-bromo-4-chloro-3-indolyl-phosphate / Nitro blue tetrazolium chloride solution, BCIP/NBT Alkaline Phosphatase Substrate solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.83
Pricing and availability is not currently available.

Product Name

BCIP®/NBT Liquid Substrate System, ready to use solution

Quality Level

form

liquid

storage temp.

2-8°C

Application

A ready-to-use, precipitating substrate system for alkaline phosphatase. This substrate system produces an insoluble NBT diformazan end product that is blue to purple in color and can be observed visually. Immunohistochemical staining with BCIP/NBT utilizes requires organic mounting media and can be counterstained with Nuclear Fast Red or Light Green. BCIP/NBT is generally a more sensitive method than Fast Red. Not recommended for ELISA (microwell) procedures.
BCIP®/NBT Liquid Substrate System has been used:
  • for staining alkaline phosphatase(ALP) in bone marrow stromal cells[1]
  • as a substrate in western blot detection of matrix metalloproteinase-2 specific antibody[2]
  • as a substrate for streptavidin-alkaline phosphatase in the enzyme-linked immunospot (ELISpot) assays[3]

Caution

It is recommended to store the product between 4 - 8 °C. Do not freeze. The product may be stored at room temperature for up to one year. If a precipitate forms, the solution can be gently swirled or filtered.

Legal Information

BCIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salma E Jiménez-Badillo et al.
Scientific reports, 7(1), 13351-13351 (2017-10-19)
The Catsper1 gene, whose expression is restricted to male germ cells, has great importance in reproductive biology because of its function in sperm motility and fertilization. We previously reported that the promoter of this gene has transcriptional activity in either
Jungho Jang et al.
Scientific reports, 8(1), 7211-7211 (2018-05-10)
The conversion of carbon dioxide to formate is a fundamental step for building C1 chemical platforms. Methylobacterium extorquens AM1 was reported to show remarkable activity converting carbon dioxide into formate. Formate dehydrogenase 1 from M. extorquens AM1 (MeFDH1) was verified
The evaluation of angiogenesis and matrix metalloproteinase-2 secretion in bone marrow of multiple myeloma patients before and after the treatment
Bolkun L, et al.
Advances in Medical Sciences, 58(1), 118-125 (2013)
Epigenetic regulation of Runx2 transcription and osteoblast differentiation by nicotinamide phosphoribosyltransferase
Ling M, et al.
Cell & Bioscience, 7(1), 27-27 (2017)
Gerry Nganou et al.
Frontiers in molecular neuroscience, 11, 234-234 (2018-07-26)
The building of the brain is a multistep process that requires the coordinate expression of thousands of genes and an intense nucleocytoplasmic transport of RNA and proteins. This transport is mediated by karyopherins that comprise importins and exportins. Here, we

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Protocols

BCIP/NBT substrate produces stable, intense color for alkaline phosphatase assays, ideal for visual observation.

BCIP/NBT substrate produces stable, intense color for alkaline phosphatase assays, ideal for visual observation.

BCIP/NBT substrate produces stable, intense color for alkaline phosphatase assays, ideal for visual observation.

BCIP/NBT substrate produces stable, intense color for alkaline phosphatase assays, ideal for visual observation.

Questions

1–2 of 2 Questions  
  1. Should this be applied to fixed cells or live cells for staining?

    1 answer
    1. Product B1911 is not formulated as a balanced salt solution. Therefore, it should be applied to fixed cells, as live cells are not suitable for staining with this solution.

      Helpful?

  2. Is the BCIP in your BCIP/NBT Liquid Substrate part number B1911 5-bromo-4-chloro-3'-indolyphosphate p-toluidine salt?

    1 answer
    1. The BCIP used in Product B1911 is not 5-bromo-4-chloro-3'-indolyphosphate p-toluidine salt (CAS# 6578-06-9).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service