Skip to Content
Merck
All Photos(4)

Documents

A9272

Sigma-Aldrich

Adenosine 3′-monophosphate

from yeast

Synonym(s):

3′-AMP, 3′-Adenylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N5O7P
CAS Number:
Molecular Weight:
347.22
Beilstein:
54478
EC Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.51

biological source

yeast

Assay

≥98% (HPLC)

form

powder

solubility

1 N NH4OH: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](OP(O)(O)=O)[C@H]3O

InChI

1S/C10H14N5O7P/c11-8-5-9(13-2-12-8)15(3-14-5)10-6(17)7(4(1-16)21-10)22-23(18,19)20/h2-4,6-7,10,16-17H,1H2,(H2,11,12,13)(H2,18,19,20)/t4-,6-,7-,10-/m1/s1

InChI key

LNQVTSROQXJCDD-KQYNXXCUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Adenosine 3′-monophosphate (3′-AMP) is a metabolite produced from hydrolysis of 2′,3′-cAMP by a family of metal-dependent phosphodiesterases. 3′-AMP inhibits proliferation of preglomerular vascular smooth muscle cells and glomerular mesangial cells via A2B receptors. 2′,3′-cAMP and 3′-AMP represent a new unexplored pathway for adenosine-based cell regulation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumit Bansal et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1011, 171-178 (2016-01-17)
Detoxification of lithocholic acid (LCA) to lithocholic acid sulfate (LCA-S) is catalyzed by sulfotransferases, mainly SULT2A1. We developed and validated an ultra-high performance liquid chromatography-tandem mass spectrometric (UPLC-MS/MS) method to quantify human liver cytosolic-dependent LCA sulfation. Chromatographic separation was achieved
Chun Yuan et al.
Molecular medicine reports, 16(4), 4529-4536 (2017-08-30)
Bone fracture healing is a complex process, which is associated with several factors, including age and osteoporosis. Certain genes and biological processes that may contribute to fracture healing, have been identified following developments in systems biology and molecular biology technologies
Aksorn Saengtienchai et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 214, 28-35 (2018-09-04)
There have been many reports regarding toxic chemicals in birds. Chemicals are mainly metabolized in the liver through phase I oxidation by cytochrome P450 (CYP) and phase II conjugation by conjugated enzymes, such as UDP-glucuronosyltransferase (UGT), sulfotransferase (SULT), glutathione-S-transferase (GST)
Tristan A Smythe et al.
Environmental science & technology, 51(12), 7245-7253 (2017-05-26)
The inhibitory effects of five novel brominated flame retardants, 1,2-bis(2,4,5-tribromophenoxy)ethane (BTBPE), decabromodiphenylethane (DBDPE), 2-ethylhexyl-2,3,4,5-tetrabromobenzoate (EH-TBB), bis(2-ethylhexyl)tetrabromophthalate (BEH-TEBP), and β-tetrabromoethylcyclohexane (β-TBECH), on thyroid hormone deiodinase (DIO) and sulfotransferase (SULT) activity were investigated using human in vitro liver microsomal and cytosolic bioassays.
Jiří Vrba et al.
Metabolites, 10(8) (2020-08-19)
Natural phenolic compounds are known to be metabolized by phase II metabolic reactions. In this study, we examined the in vitro sulfation of the main constituents of silymarin, an herbal remedy produced from the fruits of the milk thistle. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service