Skip to Content
Merck
All Photos(1)

Documents

I4625

Sigma-Aldrich

Inosine 5′-monophosphate disodium salt hydrate

from yeast, Grade III, ≥98% (HPLC)

Synonym(s):

5′-IMP-Na2, 5′-Inosinic acid disodium salt hydrate, I-5′-P, IMP, Inosinic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N4O8PNa2 · xH2O
CAS Number:
Molecular Weight:
392.17 (anhydrous basis)
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

yeast

type

Grade III

Assay

≥98% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)n2cnc3C(=O)NC=Nc23

InChI

1S/C10H13N4O8P.2Na.H2O/c15-6-4(1-21-23(18,19)20)22-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;;;/h2-4,6-7,10,15-16H,1H2,(H,11,12,17)(H2,18,19,20);;;1H2/q;2*+1;/p-2/t4-,6-,7-,10-;;;/m1.../s1

InChI key

KQCIGEIXDXQSGM-MSQVLRTGSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Inosine 5′-monophosphate (IMP) is used as a substrate to study the distribution, specificity and kinetics of inosine-5′-monophosphate dehydrogenase (IMPDH).
Inosine 5′-monophosphate disodium salt hydrate has been used:
  • as a 5′-ribonucleotide tastant compound in taste-detection threshold tests in chicks
  • for standard curve generation in high-performance liquid chromatography (HPLC) to quantify IMP from prawn tail tissue samples
  • as a component of McCoy′s 5A complete medium for Plasmodium vivax culture

Biochem/physiol Actions

Inosine 5′-monophosphate (IMP) is a dietary nucleotide that is synthesized from adenosine monophosphate (AMP) in eukaryotes. It is a food additive and is essential for intestinal health and immunity.
Inosine monophosphate, or inosinic acid, is the ribonucleotide of hypoxanthine and the first nucleotide formed during purine synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shubo Li et al.
Food chemistry, 216, 275-281 (2016-09-07)
5'-adenylic acid deaminase (AMP deaminase), an important enzyme for the food industry, can catalyze the irreversible hydrolysis of adenosine monophosphate (AMP) to inosine monophosphate (IMP) and ammonia. In this study, a new strain was screened that efficiently produces 3191.6U/g of
G Q Zhang et al.
Poultry science, 87(7), 1364-1369 (2008-06-26)
An experiment was conducted to evaluate the effects of dietary inosinic acid (inosine 5'-monophosphate, IMP) on carcass characteristics, meat quality, and deposition of IMP in Arbor Acres broilers. A total of two hundred forty 1-d-old male Arbor Acres broilers were
Shira L Cheled-Shoval et al.
Poultry science, 96(7), 2206-2218 (2017-03-25)
The sense of taste has a key role in nutrient sensing and food intake in animals. A standardized and simple method for determination of tastant-detection thresholds is required for chemosensory research in poultry. We established a 24-h, 2-alternative, forced-choice solution-consumption
Effect of salt treatments on survival and consumer acceptability of freshwater prawn, Macrobranchium rosenbergii
Ciaramella MA, et al.
Aquaculture (Amsterdam, Netherlands), 428 (428)
Bjarne G Hansen et al.
BMC microbiology, 11, 202-202 (2011-09-20)
Many secondary metabolites produced by filamentous fungi have potent biological activities, to which the producer organism must be resistant. An example of pharmaceutical interest is mycophenolic acid (MPA), an immunosuppressant molecule produced by several Penicillium species. The target of MPA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service