Skip to Content
Merck
All Photos(5)

Key Documents

A1752

Sigma-Aldrich

Adenosine 5′-monophosphate sodium salt

from yeast, ≥99%

Synonym(s):

5′-Adenylic acid, A-5′-P, AMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N5O7P · xNa+ · yH2O
CAS Number:
Molecular Weight:
347.22 (anhydrous free acid basis)
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.51

biological source

yeast

Quality Level

Assay

≥99%

form

powder

solubility

H2O: soluble

storage temp.

−20°C

SMILES string

NC1=NC=NC2=C1N=CN2[C@H]3[C@H](O)[C@H](O)[C@]((COP(O)(O)=O)([H])O3.C.

Looking for similar products? Visit Product Comparison Guide

Application

Adenosine 5′-monophosphate sodium salt has been used:
  • as a component of reactivation medium to reactivate mouse sperm extracted from caudae epididymides for sperm motility studies
  • as a substrate in signal amplification by reversible exchange (SABRE) method to develop biocompatible SABRE hyperpolarization
  • as a substrate in wheat and Aspergillus niger phytase, assay
  • in luciferase assay to monitor intracellular α-synuclein oligomerization in S1S2 cells
  • to study its effects on induction of therapeutic hypothermia and neuronal damage in a rat model of cardiac arrest (CA) and resuscitation

Biochem/physiol Actions

Adenosine 5′-monophosphate (5′-AMP) is used to enhance flavor in meat and poultry soups. It is also used as a food additive for specific nutritional purposes. 5′-AMP is an activator of a class of protein kinases known as AMP-activated protein kinase (AMPK). Along with inosine monophosphate (IMP), AMP mediates the dissociation of skeletal muscle actomyosin to actin and myosin. Therefore, these two factors may be involved in causing the resolution of rigor mortis in muscles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexey S Kiryutin et al.
Magnetic resonance in chemistry : MRC (2021-06-05)
Signal amplification by reversible exchange (SABRE) is a popular method for generating strong signal enhancements in nuclear magnetic resonance (NMR). In SABRE experiments, the source of polarization is provided by the nonthermal spin order of parahydrogen (pH2 , the H2
D Grahame Hardie
Genes & development, 25(18), 1895-1908 (2011-09-23)
AMP-activated protein kinase (AMPK) is a sensor of energy status that maintains cellular energy homeostasis. It arose very early during eukaryotic evolution, and its ancestral role may have been in the response to starvation. Recent work shows that the kinase
The oxygen isotope composition of phosphate released from phytic acid by the activity of wheat and Aspergillus niger phytase
von Sperber C, et al.
Biogeosciences (Online), 12(13), 4175-4184 (2015)
Akihiro Okitani et al.
Bioscience, biotechnology, and biochemistry, 72(8), 2005-2011 (2008-08-08)
We investigated to determine why heating of squid muscle at 60 degrees C induced the liberation of actin from myofibrils. When a mixture of a myofibrillar fraction and a low-molecular sarcoplasmic fraction prepared from squid muscle was heated at 60
Daoying Wang et al.
Journal of the science of food and agriculture, 96(5), 1467-1473 (2015-05-09)
Adenosine 5'-monophosphate (AMP) is often used in meat and poultry soups as a flavor enhancer (flavor modifier), or as food additives for specific nutritional purposes. Our previous research as well as evidence from others showed that actomyosin could be dissociated

Protocols

To measure 5′-nucleotidase activity, this procedure uses adenosine 5’-monophosphate and a color reagent to create a standard curve for determining the micromoles of phosphorus liberated.

To measure 5′-nucleotidase activity, this procedure uses adenosine 5’-monophosphate and a color reagent to create a standard curve for determining the micromoles of phosphorus liberated.

To measure 5′-nucleotidase activity, this procedure uses adenosine 5’-monophosphate and a color reagent to create a standard curve for determining the micromoles of phosphorus liberated.

To measure 5′-nucleotidase activity, this procedure uses adenosine 5’-monophosphate and a color reagent to create a standard curve for determining the micromoles of phosphorus liberated.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service