Skip to Content
Merck
All Photos(4)

Key Documents

A0386

Sigma-Aldrich

Adenosine 3′-monophosphate sodium salt

from yeast

Synonym(s):

3′-AMP, 3′-Adenylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N5O7P
CAS Number:
Molecular Weight:
347.22
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

yeast

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

water: 100 mg/mL, clear to very slightly hazy, colorless to very faintly yellow

storage temp.

−20°C

SMILES string

[Na].Nc1ncnc2n(cnc12)C3OC(CO)C(OP(O)(O)=O)C3O

InChI

1S/C10H14N5O7P.Na.H/c11-8-5-9(13-2-12-8)15(3-14-5)10-6(17)7(4(1-16)21-10)22-23(18,19)20;;/h2-4,6-7,10,16-17H,1H2,(H2,11,12,13)(H2,18,19,20);;

InChI key

SHIVFKVYHYZMPS-UHFFFAOYSA-N

General description

Adenosine 3′-monophosphate (3′-AMP) is a metabolite produced from the hydrolysis of 2′,3′-cAMP by a family of metal-dependent phosphodiesterases. 3′-AMP inhibits the proliferation of preglomerular vascular smooth muscle cells and glomerular mesangial cells via A2B receptors. 2′,3′-cAMP, and 3′-AMP represent a new unexplored pathway for adenosine-based cell regulation.

Application

Adenosine 3′-monophosphate sodium salt has been used as a marker in thin-layer chromatography (TLC).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Martinez et al.
The Journal of biological chemistry, 275(31), 24222-24230 (2000-05-10)
We have previously identified a HeLa cell 3' exonuclease specific for degrading poly(A) tails of mRNAs. Here we report on the purification and identification of a calf thymus 54-kDa polypeptide associated with a similar 3' exonuclease activity. The 54-kDa polypeptide
2′-AMP and 3′-AMP inhibit proliferation of preglomerular vascular smooth muscle cells and glomerular mesangial cells via A2B receptors.
Jackson EK, Gillespie DG, Dubey RK.
Journal of Pharmacology and Experimental Therapeutics, 337, 4440450-4440450 (2011)
Feng Rao et al.
Biochemical and biophysical research communications, 398(3), 500-505 (2010-07-06)
The recent report of 2',3'-cAMP isolated from rat kidney is the first proof of its biological existence, which revived interest in this mysterious molecule. 2',3'-cAMP serves as an extracellular adenosine source, but how it is degraded remains unclear. Here, we
Edwin K Jackson et al.
Hypertension (Dallas, Tex. : 1979), 56(1), 151-158 (2010-06-03)
Recently we discovered that intact kidneys release into the extracellular compartment 2',3'-cAMP (a positional isomer of 3',5'-cAMP with unknown pharmacology) and metabolize 2',3'-cAMP to 2'-AMP, 3'-AMP, and adenosine. Because adenosine inhibits growth of vascular smooth muscle cells and mesangial cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service