Skip to Content
Merck
All Photos(2)

Key Documents

A7791

Sigma-Aldrich

N-Acetyl-D-lactosamine

≥98% (TLC)

Synonym(s):

β-D-Gal-(1→4)-D-GlcNAc, 2-Acetamido-2-deoxy-4-O-β-D-galactopyranosyl-D-glucopyranose, LN, N-Acetyl-4-O-(β-D-galactopyranosyl)-D-glucosamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H25NO11
CAS Number:
Molecular Weight:
383.35
Beilstein:
96808
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (TLC)

form

powder

optical activity

[α]25/D 27.5 to 33.5 °, c = 0.5% (w/v) in water

technique(s)

thin layer chromatography (TLC): suitable

impurities

≤8% water

color

white

mp

170 -185  °C ((338 - 365 °F ))

solubility

water: 50 mg/mL, clear to very slightly hazy, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)[C@@H]1O

InChI

1S/C14H25NO11/c1-4(18)15-7-9(20)12(6(3-17)24-13(7)23)26-14-11(22)10(21)8(19)5(2-16)25-14/h5-14,16-17,19-23H,2-3H2,1H3,(H,15,18)/t5-,6-,7-,8+,9-,10+,11-,12-,13?,14+/m1/s1

InChI key

KFEUJDWYNGMDBV-RPHKZZMBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Acetyl-D-lactosamine is used as a specific lectin target molecule in the identification and differentiation of sugar binding molecules such as the galectins. N-Acetyl-D-lactosamine is used in studies of galactosidase, fucosyltransferase, sialyltransferase, and lectin inhibition.
Useful in studies of galactosidase, fucosyltransferase, sialyltransferase, and lectin inhibition.

Preparation Note

Synthetic

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph Darbellay et al.
EBioMedicine, 25, 73-86 (2017-11-04)
Outcomes of Zika virus (ZIKV) infection in pregnant women vary from the birth of asymptomatic offspring to abnormal development and severe brain lesions in fetuses and infants. There are concerns that offspring affected in utero and born without apparent symptoms
Magdalena Osiewicz et al.
Journal of clinical medicine, 8(7) (2019-07-03)
The aim was to determine the occurrence of temporomandibular disorders (TMDs) in patients with Lyme disease (LD), and to estimate the contribution of factors that may identify TMD among LD patients. In seventy-six (N = 76) adult patients with LD
Vanessa Dumeaux et al.
PLoS computational biology, 13(9), e1005680-e1005680 (2017-09-29)
Although systemic immunity is critical to the process of tumor rejection, cancer research has largely focused on immune cells in the tumor microenvironment. To understand molecular changes in the patient systemic response (SR) to the presence of BC, we profiled
Moritz Augustin et al.
PLoS computational biology, 13(6), e1005545-e1005545 (2017-06-24)
The spiking activity of single neurons can be well described by a nonlinear integrate-and-fire model that includes somatic adaptation. When exposed to fluctuating inputs sparsely coupled populations of these model neurons exhibit stochastic collective dynamics that can be effectively characterized
Paulina Sindrewicz et al.
Scientific reports, 9(1), 11851-11851 (2019-08-16)
Galectins are involved in the regulation of divergent physiological and pathological processes and are increasingly recognized to play important roles in a number of diseases. However, a simple and effective way in assessing galectin-ligand interactions is lacking. Our examination of

Articles

O-Linked glycoproteins are usually large proteins with a molecular mass of >200 kDa. Glycosylation generally occurs in high-density clusters and may represent as much as 50-80% of the overall mass.

O-Linked glycoproteins are usually large proteins with a molecular mass of >200 kDa. Glycosylation generally occurs in high-density clusters and may represent as much as 50-80% of the overall mass.

O-Linked glycoproteins are usually large proteins with a molecular mass of >200 kDa. Glycosylation generally occurs in high-density clusters and may represent as much as 50-80% of the overall mass.

O-Linked glycoproteins are usually large proteins with a molecular mass of >200 kDa. Glycosylation generally occurs in high-density clusters and may represent as much as 50-80% of the overall mass.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service