Skip to Content
Merck
All Photos(4)

Documents

F3004

Sigma-Aldrich

Fetuin from fetal bovine serum

lyophilized powder

Synonym(s):

Fetuin from fetal calf serum

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

bovine serum (fetal)

form

lyophilized powder

technique(s)

HPLC: suitable
MALDI-MS: suitable

impurities

~0.2% free N-acetylneuraminic acid

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... FETUB(504615)

Looking for similar products? Visit Product Comparison Guide

General description

Fetuin is an abundant glycosylated protein from fetal bovine serum and functions as a transport and storage protein. It is also called the α2-Heremans-Schmid glycoprotein (AHSG) and corresponds to a molecular weight of 64 kDa. It belongs to the statin family and is synthesized principally in the liver and partially in the kidneys, placenta and the tongue.

Application

Fetuin from fetal bovine serum has been used:
  • as a standard glycoprotein in matrix-assisted laser desorption/ionization-mass spectrometry (MALDI-MS) and high-performance liquid chromatography-fluorescence detection analysis (HPLC-FLD)
  • as a ligand in carbohydrate binding assay of pertussis toxin vaccine
  • in the synthesis of fetuin-A conjugated gold nanoparticles (F-GNPs) for drug carrier studies

Biochem/physiol Actions

In humans, polymorphism in the fetuin gene is implicated in type 2 diabetes mellitus and modulates adipocyte functionality. Elevated level of fetuin is observed in obesity and nonalcoholic fatty liver diseases (NAFLD). Fetuin is a potential marker in clinical diagnosis of metabolic disorders.

Preparation Note

Further processing of F2379 by gel filtration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of an in vitro Biochemical Assay System for the Measurement of Residual Toxin Activities in Pertussis Toxin Containing Vaccines
Ting Yuen C, et al.
Journal of vaccines & vaccination, 08 (2017)
M Demetriou et al.
The Journal of biological chemistry, 271(22), 12755-12761 (1996-05-31)
The serum glycoprotein fetuin is expressed during embryogenesis in multiple tissues including limb buds and has been shown to promote bone remodeling and stimulate cell proliferation in vitro. In this report, we demonstrate that fetuin antagonizes the antiproliferative action of
Fetuin-A (alpha2HS-glycoprotein) is a major serum adhesive protein that mediates growth signaling in breast tumor cells
Sakwe AM, et al.
Test, 285(53), 41827-41835 (2010)
Iqra Munir et al.
International journal of biological macromolecules, 101, 131-145 (2017-03-23)
Present study was conducted to establish the interaction of bovine fetuin-A to validate its binding modalities with doxorubicin (Dox). Fetuin-A was purified to highest purity and monodispersity. Green synthesis of fetuin-A conjugated gold nanoparticles (F-GNPs) has been performed giving typical
Fast purification of glycans and glycopeptides using silk-packed micropipette tip for matrix-assisted laser desorption/ionization-mass spectrometry and high-performance liquid chromatography-fluorescence detection analysis
Kayili H, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 139(2), 492-499 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service