Skip to Content
Merck
All Photos(1)

Key Documents

A2795

Sigma-Aldrich

N-Acetyl-D-galactosamine

≥98% (HPLC)

Synonym(s):

2-Acetamido-2-deoxy-D-galactose, D-GalNAc, N-Acetylchondrosamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO6
CAS Number:
Molecular Weight:
221.21
Beilstein:
2331340
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white

mp

160 °C

solubility

water: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O

InChI

1S/C8H15NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-8,10,12-14H,2H2,1H3,(H,9,11)/t4-,5-,6+,7-,8?/m1/s1

InChI key

OVRNDRQMDRJTHS-KEWYIRBNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Acetyl-D-galactosamine (GalNAc), an amino sugar, is a component of many O-linked and N-linked glycan structures. As uridine diphosphate (UDP)-GalNAc, GalNAc is the initial O-linked sugar to many serine and threonine residues in protein glycosylations.

Application

N-Acetyl-D-galactosamine has been used:
  • as a Dolichos biflorus agglutinin (DBA) haptenic sugar in lectin bead binding assay
  • to assess the specificity of lectin binding using lectin blot inhibition
  • in immunohistochemistry to pre-adsorb Wisteria floribunda lectin

N-Acetyl-D-galactosamine (GalNAc), an aminosugar, is a component of many O-linked and N-linked glycan structures. As UDP-GalNAc, GalNAc is the intial O-linked sugar to many serine and threonine residues in protein glycosylations.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dezemon Zingue et al.
Scientific reports, 8(1), 6778-6778 (2018-05-02)
Contaminations and fastidiousness of M. ulcerans may have both hamper isolation of strains from environmental sources. We aimed to optimize decontamination and culture of environmental samples to circumvent both limitations. Three strains of M. ulcerans cultured onto Middlebrook 7H10 at
Ju Huang et al.
Molecular therapy. Methods & clinical development, 5, 241-258 (2017-06-13)
Fabry disease is a rare lysosomal storage disorder (LSD). We designed multiple recombinant lentivirus vectors (LVs) and tested their ability to engineer expression of human α-galactosidase A (α-gal A) in transduced Fabry patient CD34
David J Gill et al.
Trends in cell biology, 21(3), 149-158 (2010-12-15)
O-GalNAc glycosylation of proteins confers essential structural, protective and signaling roles in eumetazoans. Addition of O-glycans onto proteins is an extremely complex process that regulates both sites of attachment and the types of oligosaccharides added. Twenty distinct polypeptide GalNAc-transferases (GalNAc-Ts)
F H M van Zelst et al.
Faraday discussions, 218(0), 219-232 (2019-05-24)
To detect and characterise compounds in complex matrices, it is often necessary to separate the compound of interest from the matrix before analysis. In our previous work, we have developed the coupling of supercritical fluid chromatography (SFC) with nuclear magnetic
Philip Steiner et al.
Frontiers in plant science, 11, 873-873 (2020-07-28)
Adaptation strategies in freezing resistance were investigated in Klebsormidium crenulatum, an early branching streptophyte green alga related to higher plants. Klebsormidium grows naturally in unfavorable environments like alpine biological soil crusts, exposed to desiccation, high irradiation and cold stress. Here

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service