Skip to Content
Merck
All Photos(3)

Documents

A8625

Sigma-Aldrich

N-Acetyl-D-glucosamine

≥99% (HPLC)

Synonym(s):

2-Acetamido-2-deoxy-D-glucose, D-GlcNAc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO6
CAS Number:
Molecular Weight:
221.21
Beilstein:
1727589
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

crab (red)

Quality Level

Assay

≥99% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white to off-white

mp

211 °C (dec.) (lit.)

solubility

H2O: 50 mg/mL, clear to very slightly hazy (colorless to faint yellow solution)

storage temp.

−20°C

SMILES string

CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

InChI

1S/C8H15NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-8,10,12-14H,2H2,1H3,(H,9,11)/t4-,5-,6-,7-,8?/m1/s1

InChI key

OVRNDRQMDRJTHS-RTRLPJTCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-acetylglucosamine (GlcNAc) is the acetylated derivative of glucosamine (GlcN). It is an important component of proteoglycans, glycoproteins, glycosaminoglycans (GAGs), and other building units found in connective tissue.

Application

N-Acetyl-D-glucosamine has been used:
  • as sugar for competitive inhibition in lectin histochemistry
  • as a component in Barbour-Stonner-Kelly (BSK) medium for cultivating Borrelia burgdorferi spirochetes strains
  • as a component of binding buffer to suspend Streptococcus pneumoniae strains for inhibition assay to examine the specificity of M-ficolin binding to S. pneumoniae strains

Biochem/physiol Actions

N-acetylglucosamine (GlcNAc) is mainly used in the food, cosmetics, and pharmaceutical industries.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mikkel Girke Jørgensen et al.
Genes & development, 27(10), 1132-1145 (2013-05-15)
Many bacterial small RNAs (sRNAs) regulate gene expression through base-pairing with mRNAs, and it has been assumed that these sRNAs act solely by this one mechanism. Here we report that the multicellular adhesive (McaS) sRNA of Escherichia coli uniquely acts
Colette Cywes-Bentley et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(24), E2209-E2218 (2013-05-30)
Microbial capsular antigens are effective vaccines but are chemically and immunologically diverse, resulting in a major barrier to their use against multiple pathogens. A β-(1→6)-linked poly-N-acetyl-d-glucosamine (PNAG) surface capsule is synthesized by four proteins encoded in genetic loci designated intercellular
Lucy G Weaver et al.
Carbohydrate research, 371, 68-76 (2013-03-26)
Poly-N-acetylglucosamine (PNAG) saccharides are an important constituent of bacterial biofilms, such as those produced by Staphylococcus aureus. We have developed a simple two-step iterative method for the synthesis of β-(1→6)-glucosamine oligosaccharides that are structurally similar to PNAG. We illustrate the
Kwang-Hyup Lee et al.
Tissue & cell, 48(4), 361-369 (2016-05-29)
The morphological characteristics and glycoconjugate composition of the vomeronasal organ (VNO) of the horse was investigated using histological, immunohistochemical, and lectin histochemical methods. The VNO is bilaterally located at the base of the nasal septum, has a tubular structure surrounded
Gerald W Hart et al.
Nature, 446(7139), 1017-1022 (2007-04-27)
All animals and plants dynamically attach and remove O-linked beta-N-acetylglucosamine (O-GlcNAc) at serine and threonine residues on myriad nuclear and cytoplasmic proteins. O-GlcNAc cycling, which is tightly regulated by the concerted actions of two highly conserved enzymes, serves as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service