Skip to Content
Merck
All Photos(1)

Documents

A7275

Sigma-Aldrich

L-2-Aminoadipic acid

≥98% (TLC), powder, glutamine synthetase inhibitor

Synonym(s):

(S)-2-Aminohexanedioic acid, L-Homoglutamic acid, Aad

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
161.16
Beilstein:
1724348
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

product name

L-2-Aminoadipic acid, ≥98% (TLC)

Assay

≥98% (TLC)

form

powder

mp

203-205 °C (dec.) (lit.)

SMILES string

N[C@@H](CCCC(O)=O)C(O)=O

InChI

1S/C6H11NO4/c7-4(6(10)11)2-1-3-5(8)9/h4H,1-3,7H2,(H,8,9)(H,10,11)/t4-/m0/s1

InChI key

OYIFNHCXNCRBQI-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-2-Aminoadipic acid/α-aminoadipate is a gliotoxin and is a structural analog of glutamine. It is generated by the catabolism of lysine saccharopine (SAC)/pipecolic acid (PIP) pathways.

Application

L-2-Aminoadipic acid has been used as a gliotoxin specific for astrocytes in rat. It has also been as an internal standard in the amino acid analysis from fermented food sample.

Biochem/physiol Actions

L-2-Aminoadipic acid inhibits glutamine synthetase and γ-glutamylcysteine synthetase. It modulates glucose metabolism and is present in higher levels in diabetic patients. L-2-Aminoadipic acid plays a key role in suppressing autophagy in C2C12 myotubes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fermentation of seeds of Teff (Eragrostis teff), grass-pea (Lathyrus sativus), and their mixtures: aspects of nutrition and food safety
Yigzaw Y, et al.
Journal of Agricultural and Food Chemistry, 52(5), 1163-1169 (2004)
Inhibition of the glutamate transporter and glial enzymes in rat striatum by the gliotoxin, ocaminoadipate
McBean, Gethin J
British Journal of Pharmacology, 113(2), 536-540 (1994)
2-Aminoadipic acid is a biomarker for diabetes risk
Wang TJ, et al.
The Journal of Clinical Investigation, 123(10), 4309-4317 (2013)
Astrocytes Protect Neurons in the Hippocampal CA3 Against Ischemia by Suppressing the Intracellular Ca2+ Overload
Sun C, et al.
Frontiers in Cellular Neuroscience, 12, 2168-2175 (2018)
GABA ergic inhibition through synergistic astrocytic neuronal interaction transiently decreases vasopressin neuronal activity during hypoosmotic challenge
Wang YF, et al.
The European Journal of Neuroscience, 37(8), 1260-1269 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service