
L-2-Aminoadipic acid

Synonym(s):
(S)-2-Aminohexanedioic acid, L-Homoglutamic acid, Aad
Linear Formula:
HO2C(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
161.16
Products
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing