Skip to Content
Merck
All Photos(2)

Documents

A7400

Sigma-Aldrich

D-2-Aminoadipic acid

Synonym(s):

(R)-2-Aminohexanedioic acid, D-Homoglutamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
161.16
Beilstein:
1724347
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

form

powder

mp

208-210 °C (lit.)

SMILES string

N[C@H](CCCC(O)=O)C(O)=O

InChI

1S/C6H11NO4/c7-4(6(10)11)2-1-3-5(8)9/h4H,1-3,7H2,(H,8,9)(H,10,11)/t4-/m1/s1

InChI key

OYIFNHCXNCRBQI-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-2-Aminoadipic acid has been used as an inhibitor of glutamine synthetase in spontaneously immortalized human cell line MIO-M1.

Biochem/physiol Actions

D-2-Aminoadipic acid is a glutamate analog. It acts as an inhibitor of glutamate synthetase. D-2-Aminoadipic acid exhibits gliotoxicity towards mitotic astrocytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wang-Yang Xu et al.
The Journal of endocrinology, 243(2), 111-123 (2019-08-28)
Obesity and type 2 diabetes (T2D) are both complicated endocrine disorders resulting from an interaction between multiple predisposing genes and environmental triggers, while diet and exercise have key influence on metabolic disorders. Previous reports demonstrated that 2-aminoadipic acid (2-AAA), an
Yang Guo et al.
Experimental eye research, 210, 108703-108703 (2021-07-20)
Diabetic retinopathy (DR) is a vision-loss complication caused by diabetes with high prevalence. During DR, the retinal microvascular injury and neurodegeneration derived from chronic hyperglycemia have attracted global attention to retinal Müller cells (RMCs), the major macroglia in the retina
D R Brown et al.
Journal of neurocytology, 27(2), 109-118 (1998-06-03)
The mechanism of action of the glutamate analogue alpha-aminoadipic (AAA) acid was investigated in terms of its toxicity to cultured astrocytes. AAA was more toxic to type 1 astrocytes than type 2 astrocytes. Also the higher toxicity of the L-isomer
T P Hicks et al.
Neuroscience research, 24(2), 139-150 (1996-01-01)
Many studies on long-term potentiation (LTP) in hippocampal region CA1 focus on receptor-mediated events that are often presumed to be linked to postsynaptic processes. Whereas it is now well-known that LTP consists of multiple components involving increases in postsynaptic responsiveness
S Servotte et al.
Protoplasma, 229(2-4), 215-220 (2006-12-21)
Small GTP-binding proteins of the Rho family (RhoA, Cdc42, Rac1) regulate the organisation and the turnover of the cell's cytoskeleton and adhesion structures. A significant function of these cellular structures is to translate and counterbalance forces applied to, or generated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service