Skip to Content
Merck
All Photos(1)

Key Documents

A4477

Sigma-Aldrich

Anti-phospho-AMPA Receptor, GluR1 Subunit (pSer845) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-phospho-AMPA Glutamate Receptor 1 (pSer845)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen 100 kDa

species reactivity

human, rat

technique(s)

dot blot: 1:1000
western blot: 1:1,000 using rat brain extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer845)

Gene Information

human ... GRIA1(2890)
rat ... Gria1(50592)

General description

Rabbit Anti-phospho-AMPA Receptor, GluR1 Subunit (pSer845) antibody recognizes human and rat AMPA Receptor, GluR1 subunit (pSer845).

Immunogen

synthetic phosphorylated peptide corresponding to a region near serine 845 of rat glutamate receptor subunit GluR1.

Application

Rabbit Anti-phospho-AMPA Receptor, GluR1 Subunit (pSer845) antibody can be used for western blot analysis at 1:1,000 using rat brain extracts.

Target description

AMPA is a transmembrane receptor for glutamate that regulates synapsis in the central nervous system. It is composed of 4 types of subunits, GluR1, 2, 3, 4.

Physical form

Solution in 10 mM HEPES buffer, pH 7.5, containing 150 mM NaCl, 100 μg/ml BSA and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodrigo Bainy Leal et al.
Molecular psychiatry, 25(3), 655-665 (2018-06-09)
Fear is a conscious state caused by exposure to real or imagined threats that trigger stress responses that affect the body and brain, particularly limbic structures. A sub-group of patients with mesial temporal lobe epilepsy related to hippocampus sclerosis (MTLE-HS)
Loukia Parisiadou et al.
Nature neuroscience, 17(3), 367-376 (2014-01-28)
Leucine-rich repeat kinase 2 (LRRK2) is enriched in the striatal projection neurons (SPNs). We found that LRRK2 negatively regulates protein kinase A (PKA) activity in the SPNs during synaptogenesis and in response to dopamine receptor Drd1 activation. LRRK2 interacted with
Kuang-Ti Chen et al.
Frontiers in behavioral neuroscience, 9, 162-162 (2015-07-08)
Sarcosine, an endogenous amino acid, is a competitive inhibitor of the type I glycine transporter and an N-methyl-d-aspartate receptor (NMDAR) coagonist. Recently, we found that sarcosine, an NMDAR enhancer, can improve depression-related behaviors in rodents and humans. This result differs
M W Lopes et al.
Translational psychiatry, 6(12), e986-e986 (2016-12-14)
Glucocorticoids (GC) released during stress response exert feedforward effects in the whole brain, but particularly in the limbic circuits that modulates cognition, emotion and behavior. GC are the most commonly prescribed anti-inflammatory and immunosuppressant medication worldwide and pharmacological GC treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service