Skip to Content
Merck
All Photos(1)

Documents

49163

Sigma-Aldrich

Glucose solution

BioUltra, for molecular biology, ~20% in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1281608
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

grade

for molecular biology

product line

BioUltra

form

liquid

concentration

~20% in H2O

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

refractive index

n20/D 1.362

pH

5.0-8.0 (25 °C)

cation traces

Al: ≤1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: ≤0.10
λ: 280 nm Amax: ≤0.10

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Gene Information

human ... PYGM(5837)

Application

D-(+)-Glucose is a common natural sugar involved in processes such as energy production, glycosylation, and formation of glycans that provide structure to cells. Glucose is involved in a detrimentatl process in cells called glycation. Glucose is used as a supplement for cell culture and in numerous cellular processes and molecular biology applications.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jelena A Stamenkovic et al.
Metabolism: clinical and experimental, 61(7), 978-985 (2012-02-07)
Nearly all mammalian cells express a set of genes known as clock genes. These regulate the circadian rhythm of cellular processes by means of negative and positive autoregulatory feedback loops of transcription and translation. Recent genomewide association studies have demonstrated
Barbara D Abbott et al.
Reproductive toxicology (Elmsford, N.Y.), 33(4), 491-505 (2011-12-14)
PPARs regulate metabolism and can be activated by environmental contaminants such as perfluorooctanoic acid (PFOA). PFOA induces neonatal mortality, developmental delay, and growth deficits in mice. Studies in genetically altered mice showed that PPARα is required for PFOA-induced developmental toxicity.
Erica Tambone et al.
BMC oral health, 21(1), 49-49 (2021-02-06)
Peri-implant mucositis and peri-implantitis are biofilm-related diseases causing major concern in oral implantology, requiring complex anti-infective procedures or implant removal. Microbial biosurfactants emerged as new anti-biofilm agents for coating implantable devices preserving biocompatibility. This study aimed to assess the efficacy
Takaharu Sasaki et al.
Cell reports, 28(1), 202-217 (2019-07-04)
Complex interactions between immune cells are an important component in the induction of obesity. Here, we show that Il2rg-/-Rag2-/- mice lacking all lymphocytes are resistant to diet-induced obesity. Transplantation of bone marrow cells from Rag2-/- mice, which lack only acquired
Magnus Dalén et al.
Journal of cardiovascular translational research, 8(6), 381-384 (2015-07-08)
Endomyocardial delivery in the setting of active left ventricular assist device (LVAD) support has rarely been studied. The objective was to establish a protocol for endomyocardial injections during LVAD support without compromising mechanical circulation. LVAD implantation was performed in four

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service