Skip to Content
Merck
All Photos(1)

Documents

05-533

Sigma-Aldrich

Anti-phospho-CaM Kinase II Antibody, α subunit, (Thr286), clone 22B1

clone 22B1, Upstate®, from mouse

Synonym(s):

Anti-Anti-CAMKA, Anti-Anti-CaMKIINalpha, Anti-Anti-CaMKIIalpha, Anti-Anti-MRD53, Anti-Anti-MRT63

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

22B1, monoclonal

species reactivity

rat

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr286)

Gene Information

human ... CAMK2A(815)

Specificity

phospho-CaM Kinase II α subunit

Immunogen

Peptide corresponding to residuues 281-294 of rat Cam Kinase II alpha-subunit; recognizes phosphorylated CaM Kinase II on Threonine 286

Application

Detect phospho-CaM Kinase II using this Anti-phospho-CaM Kinase II Antibody, α subunit, (Thr286), clone 22B1 validated for use in WB.

Quality

routinely evaluated by immunoblot on rat brain microsomal protein preparation

Target description

50kDa

Physical form

Format: Purified

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su-Ying Cui et al.
Journal of neurochemistry, 136(3), 609-619 (2015-11-13)
The Ca(2+) modulation in the dorsal raphe nucleus (DRN) plays an important role in sleep-wake regulation. Calmodulin-dependent kinase II (CaMKII) is an important signal-transducing molecule that is activated by Ca(2+) . This study investigated the effects of intracellular Ca(2+) /CaMKII
B L Patton et al.
Molecular biology of the cell, 4(2), 159-172 (1993-02-01)
We have visualized the distribution of autophosphorylated type II CaM kinase in neural tissue with the use of two complementary antibodies: a monoclonal antibody that binds to the alpha and beta subunits of the kinase only when they are autophosphorylated
Comparative biology of cystic fibrosis animal models.
Fisher, JT; Zhang, Y; Engelhardt, JF
Methods in Molecular Biology null
Mar Cuadrado-Tejedor et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 42(2), 524-539 (2016-08-24)
The targeting of two independent but synergistic enzymatic activities, histone deacetylases (HDACs, class I and HDAC6) and phosphodiesterase 5 (PDE5), has recently been validated as a potentially novel therapeutic approach for Alzheimer's disease (AD). Here we report the discovery of
Overexpression of calbindin-D28K in hippocampal progenitor cells increases neuronal differentiation and neurite outgrowth.
Kim, JH; Lee, JA; Song, YM; Park, CH; Hwang, SJ; Kim, YS; Kaang, BK; Son, H
Faseb Journal null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service