Skip to Content
Merck
All Photos(4)

Key Documents

A3286

Sigma-Aldrich

N-Acetyl-D-glucosamine

BioReagent, suitable for cell culture

Synonym(s):

2-Acetamido-2-deoxy-D-glucose, D-GlcNAc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO6
CAS Number:
Molecular Weight:
221.21
Beilstein:
1727589
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.75

product line

BioReagent

Assay

≥95% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

mp

211 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

InChI

1S/C8H15NO6/c1-3(11)9-5-7(13)6(12)4(2-10)15-8(5)14/h4-8,10,12-14H,2H2,1H3,(H,9,11)/t4-,5-,6-,7-,8?/m1/s1

InChI key

OVRNDRQMDRJTHS-RTRLPJTCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Acetylglucosamine (GlcNAc), a derivative of glucose is a monosaccharide. It is a white powder and is mildly sweet. It melts at 221°C. GlcNAc is 25% soluble in water. It is present in bromelain, ricin agglutinin and abrus agglutinin, in plants. In humans, it is present in tissue plasminogen activator, growth factors and hormones.

Application

N-Acetyl-D-glucosamine addition has been used:
  • as a substrate to develop dot-blots
  • as a substrate to reveal alkaline phosphatase in the immunohistochemical analysis
  • to detect specific immune complexes in western blot analysis

Biochem/physiol Actions

N-Acetylglucosamine (GlcNAc) oligomer may have the ability to initiate factors for canine polymorphonuclear cells (PMN) in vivo. It possesses wound healing and chemotactic activity. GlcNAc and its derivatives are usually employed in preparing dietary supplements and also used in therapeutic development.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucose metabolism and hexosamine pathway regulate oncogene-induced senescence
Gitenay D, et al.
Cell Death & Disease, 5(2), e1089-e1089 (2014)
Cumulus expansion, nuclear maturation and connexin 43, cyclooxygenase-2 and FSH receptor mRNA expression in equine cumulus-oocyte complexes cultured in vitro in the presence of FSH and precursors for hyaluronic acid synthesis
Dell'Aquila M E, et al.
Reproductive Biology and Endocrinology, 2(1), 44-44 (2004)
Effect of N-acetyl-d-glucosamine and d-glucosamine oligomers on canine polymorphonuclear cells in vitro
Usami Y, et al.
Carbohydrate Polymers, 36(2-3), 137-141 (1998)
Glycoprofiling effects of media additives on IgG produced by CHO cells in fed-batch bioreactors
Kildegaard H F, et al.
Biotechnology and Bioengineering, 113(2), 359-366 (2016)
Mikkel Girke Jørgensen et al.
Genes & development, 27(10), 1132-1145 (2013-05-15)
Many bacterial small RNAs (sRNAs) regulate gene expression through base-pairing with mRNAs, and it has been assumed that these sRNAs act solely by this one mechanism. Here we report that the multicellular adhesive (McaS) sRNA of Escherichia coli uniquely acts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service