Skip to Content
Merck
All Photos(5)

Documents

T8441

Sigma-Aldrich

L-Threonine

99.0-101.0%, suitable for cell culture, BioXtra, non-animal source

Synonym(s):

(2S,3R)-2-Amino-3-hydroxybutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(NH2)CO2H
CAS Number:
Molecular Weight:
119.12
Beilstein:
1721646
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%

biological source

non-animal source

Agency

USP/NF
meets EP testing specifications
meets JP testing specifications
meets USP testing specifications

product line

BioXtra

Assay

99.0-101.0%

form

powder

quality

meets EP, JP, USP testing specifications

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white

mp

256 °C (dec.) (lit.)

solubility

H2O: 1 g/10 mL, clear, colorless

application(s)

pharmaceutical (small molecule)

SMILES string

C[C@@H](O)[C@H](N)C(O)=O

InChI

1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3+/m1/s1

InChI key

AYFVYJQAPQTCCC-GBXIJSLDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Threonine is a naturally occurring amino acid belonging to the aspartate family of amino acids.

Application

L-Threonine has been used for the following :
  • To study the effect of heat on homoserine kinase activity
  • In Plackett–Burman analysis and then to study the effect of selected amino acids on monoclonal antibody production in CHO cells
  • As a substrate to determine the L-threonine dehydrogenase activity
  • To study the effect of L-Threonine on the growth rate of P. putida, to check for its tolerance towards L-Threonine

Biochem/physiol Actions

L-Threonine is considered to be an essential amino acid. L-Threonine is involved in the synthesis of mucin, a protein supporting intestinal function and integrity. L-Threonine is required for the performance of the immune system. It is also essential for O-linked glycosylation, protein phosphorylation and glycine synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Homoserine Kinase from Escherichia coli K-12: Properties,
Inhibition by L-Threonine, and Regulation of Biosynthesis
J. THEZE
Journal of Bacteriology, 118(2), 577-581 (1974)
Regulation of L-Threonine Dehydrogenase in Somatic Cell Reprogramming
Chuanchun Han
Stem Cells, 31(5) (2013)
Systems metabolic engineering of Escherichia coli for L?threonine production.
Lee HK, et al.
Molecular Systems Biology, 3(1), 149-149 (2007)
Use of a Plackett-Burman statistical design to determine the effect of selected amino acids on monoclonal antibody production in CHO cells.
Gonzalez-Leal IJ
Biotechnol. Progress, 27(6), 1709-1717 (2011)
Genome-wide identification of tolerance mechanisms toward p-coumaric acid in Pseudomonas putida
Patricia Calero
Biotechnology and Bioengineering, 115(3), 762-774 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service