Skip to Content
Merck
All Photos(3)

Key Documents

M1381

Sigma-Aldrich

4-Methylumbelliferone

≥98%

Synonym(s):

β-Methylumbelliferone, 4-MU, 7-Hydroxy-4-methylcoumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O3
CAS Number:
Molecular Weight:
176.17
Beilstein:
142217
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥98%

color

off-white to yellow

fluorescence

λex 372 nm; λem 445 nm in ethanol

SMILES string

CC1=CC(=O)Oc2cc(O)ccc12

InChI

1S/C10H8O3/c1-6-4-10(12)13-9-5-7(11)2-3-8(6)9/h2-5,11H,1H3

InChI key

HSHNITRMYYLLCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methylumbelliferone is primarily used in synthesizing medicinal compounds and as a building block for fluorescent probes. Applications include synthesis of:
  • coumarin triazole derivatives as potential antimicrobial agents.[1]
  • coumarin salen-based fluorescence sensors for Mg2+ detection.[2]
  • pyranocoumarin derivatives as anti-hyperglycemic and anti-dyslipidemic agents.[3]
  • coumarin piperazine derivatives as potential multireceptor atypical antipsychotics.[4]
  • 4-methylumbelliferyl T-antigen as a substrate for endo-α-N-acetylgalactosaminidase. [5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyranocoumarins: A new class of anti-hyperglycemic and anti-dyslipidemic agents.
Kumar A, et al.
Bioorganic & Medicinal Chemistry Letters, 19(22), 6447-6451 (2009)
Tz-Ching Yeh et al.
Freshwater biology, 65(11), 1973-1988 (2020-12-09)
Bamboo, as a pioneer vegetation, often forms forests on bare lands after catastrophic landslides. Compared to evergreen forest soil, bamboo forest soil is much more labile, with a higher percentage of microbially derived organic carbon (OC), lower molecular weight, and
Na+ triggered fluorescence sensors for Mg2+ detection based on a coumarin salen moiety.
Dong Y, et al.
Organic Letters, 13(9), 2252-2255 (2011)
Synthesis and evaluation of a class of new coumarin triazole derivatives as potential antimicrobial agents.
Shi Y and Zhou C-H
Bioorganic & Medicinal Chemistry Letters, 21(3), 956-960 (2011)
Yin Chen et al.
Journal of medicinal chemistry, 56(11), 4671-4690 (2013-05-17)
The discovery and synthesis of potential and novel antipsychotic coumarin derivatives, associated with potent dopamine D2, D3, and serotonin 5-HT1A and 5-HT2A receptor properties, are the focus of the present article. The most-promising derivative was 7-(4-(4-(6-fluorobenzo[d]isoxazol-3-yl)-piperidin-1-yl)butoxy)-4-methyl-8-chloro-2H-chromen-2-one (17m). This derivative possesses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service