Skip to Content
Merck
All Photos(1)

Key Documents

75958

Sigma-Aldrich

mono-Butyl phthalate

97.0-103.0% (T)

Synonym(s):

MBP, Phthalic acid butyl ester, mono-n-Butyl phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O4
CAS Number:
Molecular Weight:
222.24
Beilstein:
2051402
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

97.0-103.0% (T)

form

crystals

SMILES string

CCCCOC(=O)c1ccccc1C(O)=O

InChI

1S/C12H14O4/c1-2-3-8-16-12(15)10-7-5-4-6-9(10)11(13)14/h4-7H,2-3,8H2,1H3,(H,13,14)

InChI key

YZBOVSFWWNVKRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonia Hernández-Díaz et al.
Environmental health perspectives, 117(2), 185-189 (2009-03-10)
Widespread human exposure to phthalates, some of which are developmental and reproductive toxicants in experimental animals, raises concerns about potential human health risks. Underappreciated sources of exposure include phthalates in the polymers coating some oral medications. The objective of this
G D Shapiro et al.
Environment international, 83, 63-71 (2015-06-24)
Studies from several countries report increases in rates of gestational diabetes mellitus (GDM) over recent decades. Exposure to environmental chemicals could contribute to this trend. To determine the associations between plasticisers and metals measured in early pregnancy with impaired glucose
S H Swan et al.
International journal of andrology, 33(2), 259-269 (2009-11-19)
Foetal exposure to antiandrogens alters androgen-sensitive development in male rodents, resulting in less male-typical behaviour. Foetal phthalate exposure is also associated with male reproductive development in humans, but neurodevelopmental outcomes have seldom been examined in relation to phthalate exposure. To
Dorien A M van Dartel et al.
Toxicology letters, 201(2), 143-151 (2011-01-05)
The embryonic stem cell test (EST) has been shown to be a promising in vitro method for the prediction of developmental toxicity. In our previous studies, we demonstrated that the implementation of gene expression analysis in the EST may further
Liangpo Liu et al.
Environment international, 42, 78-83 (2011-04-29)
Phthalates are suspected of having adverse effects on androgen-regulated reproductive development in animals and may be toxic for human sperm. The purposes of our study were to investigate the general exposure of a Chinese reproductive age cohort to these ubiquitous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service