Skip to Content
Merck
All Photos(2)

Key Documents

441147

Sigma-Aldrich

Polystyrene

average Mw ~350,000, average Mn ~170,000

Synonym(s):

PS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

pellets

melt index

2.0-4.0 g/10 min (200°C/5kg)

mol wt

average Mn ~170,000
average Mw ~350,000

impact strength

21 J/m (Izod, ASTM D 256, 23 °C, notched)

impurities

≤5% mineral oil

transition temp

Tg 95 °C

density

1.04 g/mL at 25 °C

elongation

ASTM D 638 - 1.5% (at break)

SMILES string

c1(ccccc1)C=C

InChI

1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2

InChI key

PPBRXRYQALVLMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Extrusion molded foamed packaging, dinnerware, bottles and toys. Injection molded packaging containers, toys and disposable cutlery.

Features and Benefits

Hard, clear moderately strong polymer, biologically inert, good dimensional stability. Susceptible to exterior (UV) degradation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Päivi Grönroos et al.
Sensors (Basel, Switzerland), 19(12) (2019-06-30)
Novel hot electron-emitting working electrodes and conventional counter electrodes were created by screen printing. Thus, low-cost disposable electrode chips for bioaffinity assays were produced to replace our older expensive electrode chips manufactured by manufacturing techniques of electronics from silicon or
Prasert Akkaramongkolporn et al.
AAPS PharmSciTech, 10(2), 641-648 (2009-05-20)
The differently sulfonated styrene-divinylbenzene cross-linked copolymer cationic exchange resins were prepared by oil-in-water polymerization and varied degrees of sulfonation. Several characteristics of the obtained resins were evaluated, i.e., Fourier transform infrared spectra, the ion-exchange capacity, microscopic morphology, size, and swelling.
Hui Xia et al.
Journal of colloid and interface science, 376(1), 322-326 (2012-03-27)
The aggregation kinetics of particles in dense polystyrene latex suspensions is studied by low-coherence fiber optic dynamic light scattering. Low-coherence fiber optic dynamic light scattering is used to measure the hydrodynamic radius of the aggregates. The aggregation kinetics data obtained
J K R Weber et al.
The Review of scientific instruments, 80(8), 083904-083904 (2009-09-04)
A single-axis acoustic levitator was constructed and used to levitate liquid and solid drops of 1-3 mm in diameter at temperatures in the range -40 to +40 degrees C. The levitator comprised (i) two acoustic transducers mounted on a rigid
Laarnie Tumolva et al.
Microscopy research and technique, 75(4), 505-512 (2011-09-16)
Morphological properties of atmospheric particles are directly related to their residence time and transport behaviors, and their deposition patterns in human respiratory systems. The projected properties of particles measured by transmission electron microscopy (TEM) were combined with the particle height

Articles

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Electrospinning technique applications discussed, emphasizing control of nanofibers and assembly into 3D architectures.

Questions

1–4 of 4 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What dimension has one pellet? I am looking for a particle between 1 - 5 mm in length and width? May do you know the density?

    1 answer
    1. Unfortunately, the particle size is not a test specification for this product.

      Helpful?

  4. What is the refractive index of product_441147 ?

    1 answer
    1. Unfortunately, the refractive index of this product is not determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service