Skip to Content
Merck
All Photos(1)

Key Documents

62305

Sigma-Aldrich

Lipase from Rhizopus oryzae

powder (fine), ~10 U/mg

Synonym(s):

Lipase from Rhizopus arrhizus, Triacylglycerol acylhydrolase, Triacylglycerol lipase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

powder (fine)

Quality Level

specific activity

~10 U/mg

mol wt

Mr ~43000

storage temp.

2-8°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lipase from Rhizopus oryzae (ROL) comprises an oxyanion hole, four N-glycosylation sites, and an active site region. It possesses N-terminal presequence and prosequence.[1]
Research Area: Cell Signaling
Rhizopus oryzae lipase (ROL) is a protein synthesized in a precursor form that includes a presequence of 26 amino acids, followed by a prosequence of 97 amino acids, which is attached to the N-terminal of a mature sequence consisting of 269 amino acids.[2]

Application

Lipase from Rhizopus oryzae has been used:
  • to test its effect on 1,2-diolein synthesis and triolein ethanolysis[3]
  • for immobilization on graphene oxide support for biocatalysis studies[4]
  • to digest triglycerides (TAG) from Chlamydomonas reinhardtii and S. cerevisiae[5]

Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.

Biochem/physiol Actions

Rhizopus oryzae lipase (ROL) has been extensively researched for its regiospecificity in biodiesel production. Due to its remarkable characteristics, including 1,3-specificity, high enantioselectivity, and stability in organic solvents, ROL has garnered significant attention for applications in the energy, food, and pharmaceutical industries.[2]
Lipase from Rhizopus oryzae (ROL) acts as a catalyst for the enzymatic biosynthesis of polyglycerol polyricinoleate through a reversal of hydrolysis.[6] ROL is useful in the industrial production of structured lipids due to its 1,3-regiospecificity functionality.[1]
Tri-, di-, and monoglycerides are hydrolyzed (in decreasing order of rate).

Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol of butyric acid per minute at pH 8.0 and 40°C (tributyrin, Cat. No. 91010 as substrate) 5000 U as described above are equivalent to ~1 U using triolein, Cat. No. 62314 as substrate, at pH 8.0 and 40°C

Other Notes

Note: When triacetin is used as substrate, the pH is 7.4. Incubation time: 60 minutes.
Catalyst for the interesterification of oils and fats[7]; For removal of interfering triglycerides in the electroimmunoassay of apolipoprotein B[8]; Racemic epoxy ester resolution through enantioselective enzymatic hydrolysis[9]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Kim et al.
Enzyme and Microbial Technology, 11, 528-528 (1989)
Rhizopus oryzae lipase, a promising industrial enzyme: Biochemical characteristics, production and biocatalytic applications
L'opez-Fern'andez J, et al.
Catalysts (Basel, Switzerland), 1277-1277 (2020)
Yeongho Kim et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(7), 1652-1657 (2018-02-01)
Understanding the unique features of triacylglycerol (TAG) metabolism in microalgae may be necessary to realize the full potential of these organisms for biofuel and biomaterial production. In the unicellular green alga Chlamydomonas reinhardtii a chloroplastic (prokaryotic) pathway has been proposed
J.A. Laffitte et al.
Indian J. Chem. B, 32, 94-94 (1993)
Electroimmunoassay of apolipoprotein B in triacylglycerol-rich serum.
P Laburre et al.
Clinical chemistry, 31(5), 787-787 (1985-05-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service