Skip to Content
Merck
All Photos(1)

Documents

62302

Sigma-Aldrich

Lipase from Candida rugosa

lyophilized, powder (fine), 15-25 U/mg

Synonym(s):

CCL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

fungus (Candida rugosa)

form

lyophilized solid
powder (fine)

quality

lyophilized

specific activity

15-25 U/mg

mol wt

Mr ~67000

storage temp.

2-8°C

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Cell signaling

Lipase is a part of the alpha/beta-hydrolase fold superfamily of enzymes and is expressed in multiple tissues, including the liver, adipocytes, small intestine, and endothelial surface.

Application

Lipase from Candida rugosa has been used:
  • for the resolution of chiral compounds and the transesterification production of biodiesel.
  • for studying the immobilization of different lipase enzymes on graphene oxide support for high-efficiency biocatalysis.

Biochem/physiol Actions

Candida rugosa lipase is known to catalyze hydrolysis reactions, especially the production of ricinoleic acid.
Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids.

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol oleic acid per minute at pH 8.0 and 40 °C (triolein, Cat. No. 62314 as substrate)

Other Notes

A highly stereospecific catalyst suitable for preparative resolution of racemic acids and alcohols; Stereoselective ester synthesis; Benzyl-alkyl transesterification under mild neutral conditions; Enantioselectivity of some lipases: Review

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Hult et al.
Indian J. Chem. B, 32, 123-123 (1993)
Lipase enzymes on graphene oxide support for high-efficiency biocatalysis
Hermanova S, et al.
Applied Materials Today, 5, 200- 208 (2016)
Biochemistry, Lipase
Pirahanchi Y and Sharma S
StatPearls [Internet] (2023)
Y. Ikushima et al.
Chemistry Letters (Jpn), 109-109 (1993)
B Cambou et al.
Biotechnology and bioengineering, 26(12), 1449-1454 (1984-12-01)
Lipase from Candida cylindracea has been found to be a highly stereospecific catalyst suitable for preparative resolution of racemic acids and alcohols. Using (R, S)-2-(p-chlorophenoxy) propionic acid (whose R isomer is a herbicide) and (R, S)-sec-butanol (a versatile synthon) as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service