Skip to Content
Merck
All Photos(1)

Key Documents

SRE0028

Sigma-Aldrich

Lipase from human pancreas

Synonym(s):

Lipase from human pancreas, Triacylglycerol acylhydrolase, Triacylglycerol lipase

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352108
NACRES:
NA.54

form

lyophilized powder

storage temp.

−20°C

General description

Pancreatic lipase fulfills a key function in dietary fat absorption by hydrolyzing triglycerides into diglycerides, and subsequently into monoglycerides and free fatty acids.

Packaging

100 units in glass bottle

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service