Skip to Content
Merck
All Photos(1)

Key Documents

534730

Sigma-Aldrich

Amano Lipase from Pseudomonas fluorescens

beige-brown, ≥20,000 U/g

Synonym(s):

Lipase, triacylglycerol, Triacylglycerol lipase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.22
Pricing and availability is not currently available.

biological source

Pseudomonas fluorescens

specific activity

≥20,000 U/g, pH 8.0, 55 °C (Optimum pH and temperature)
≥20,000 U/g

color

beige-brown

InChI

1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;

InChI key

QWZUIMCIEOCSJF-CHHCPSLASA-N

Related Categories

Application

Amano Lipase from Pseudomonas fluorescens can be used as a biocatalyst in the:
  • Hydrolysis and transesterification of parabens (para-hydroxybenzoic acid esters).[1]
  • Enzymatic kinetic resolution of chlorohydrins.[2]
  • Regio-selective acylation of andrographolide with different acylating agents to synthesize andrographolide derivatives.[3]

Amano Lipase can also immobilized on silk fibroin spheres to prepare optically active epoxides by enzymatic kinetic resolution of halohydrins.[4]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immobilization of Amano lipase from Pseudomonas fluorescens on silk fibroin spheres: an alternative protocol for the enantioselective synthesis of halohydrins
Ferreira IM, et al.
Royal Society of Chemistry Advances, 7(21), 12650-12658 (2017)
Lei Wang et al.
Journal of hazardous materials, 354, 145-152 (2018-05-12)
Hydrolysis is generally considered as the main pathway for the degradation of ester-type pollutants in aquatic environments. In this study, we found that when methanol or ethanol presented as the external carbon in the activated sludge, transesterification is very important
Lipase catalyzed synthesis of antimicrobial andrographolide derivatives
Patil Harshal S, et al.
Data in Brief, 18, 1134-1141 (2018)
Highly enantioselective acylation of chlorohydrins using Amano AK lipase from P. fluorescens immobilized on silk fibroin-alginate spheres
Ferreira IM, et al.
Tetrahedron Letters, 55(36), 5062-5065 (2014)
Dominik Koszelewski et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(43), 10156-10164 (2019-05-29)
A new protocol based on lipase-catalyzed tandem reaction toward α,β-enones/enoesters is presented. For the synthesis of the desired products the tandem process based on enzyme-catalyzed hydrolysis and Knoevenagel reaction starting from enol acetates and aldehyde is developed. The relevant impact

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Hello, I am writing to confirm that this product is Lipase AK since I did not find "AK" in the title. Thank you.

    1 answer
    1. Yes, this Amano Lipase is the AK version.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service