Skip to Content
Merck
All Photos(1)

Key Documents

50637

Sigma-Aldrich

Decanoyl-L-carnitine

≥94.0% (HPLC)

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxodecyl)oxy]-1-propanaminium inner salt, L-Carnitine decanoyl ester, C10-Carnitine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H33NO4
CAS Number:
Molecular Weight:
315.45
Beilstein:
4145305
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥94.0% (HPLC)

form

powder or crystals

optical activity

[α]/D -15±3°, c = 1 in H2O

impurities

≤10% water

functional group

ester

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(CCCCCCCCC)=O)CC([O-])=O

InChI

1S/C17H33NO4/c1-5-6-7-8-9-10-11-12-17(21)22-15(13-16(19)20)14-18(2,3)4/h15H,5-14H2,1-4H3/t15-/m1/s1

InChI key

LZOSYCMHQXPBFU-OAHLLOKOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Decanoylcarnitine is useful in the diagnosis of fatty acid oxidation disorders and differentiation between biochemical phenotypes of medium-chain acyl-CoA dehydrogenase (MCAD) deficiency deficiencyoxidation disorders.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah K Davies et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(29), 10761-10766 (2014-07-09)
Sleep restriction and circadian clock disruption are associated with metabolic disorders such as obesity, insulin resistance, and diabetes. The metabolic pathways involved in human sleep, however, have yet to be investigated with the use of a metabolomics approach. Here we
Junjie Zhang et al.
Journal of separation science, 39(2), 247-255 (2015-11-01)
Rice is one of the most important food crops in the world. Metabolite composition in rice seeds varies significantly depending on genetic variety, climatic alternation and agricultural practice. Metabolomics is a powerful tool to reveal the metabolic response of rice
Fotouh R Mansour et al.
Biomedical chromatography : BMC, 27(10), 1339-1353 (2013-09-06)
Carnitine and its acylesters are a family of compounds that can be used in the early diagnosis of many diseases. Carnitine and acylcarnitines have a crucial role in fatty acid transportation. The increased level of free carnitine, total carnitine, or
Christoph Nowak et al.
Scientific reports, 8(1), 8691-8691 (2018-06-08)
Insulin resistance (IR) predisposes to type 2 diabetes and cardiovascular disease but its causes are incompletely understood. Metabolic challenges like the oral glucose tolerance test (OGTT) can reveal pathogenic mechanisms. We aimed to discover associations of IR with metabolite trajectories
Judit Bene et al.
World journal of gastroenterology, 11(42), 6671-6675 (2006-01-21)
To determine the fasting plasma carnitine ester in patients with celiac disease. We determined the fasting plasma carnitine ester profile using ESI triple quadrupol mass spectrometry in 33 adult patients with biopsy-confirmed maturity onset celiac disease maintained on long term

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service