Skip to Content
Merck
All Photos(1)

Key Documents

06206

Supelco

Octanoyl-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxooctyl)oxy]-1-propanaminium inner salt, L-Carnitine octanoyl ester, C8-Carnitine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H29NO4
CAS Number:
Molecular Weight:
287.40
Beilstein:
5955761
UNSPSC Code:
41116107
NACRES:
NB.21
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

optical activity

[α]/D -17±2°, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

impurities

≤10% water

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

C[N+](C)(C)C[C@H](OC(CCCCCCC)=O)CC([O-])=O

InChI

1S/C15H29NO4/c1-5-6-7-8-9-10-15(19)20-13(11-14(17)18)12-16(2,3)4/h13H,5-12H2,1-4H3/t13-/m1/s1

InChI key

CXTATJFJDMJMIY-CYBMUJFWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Octanoyl-L-carnitine is a medium-chain endogenous ester of L-carnitine,[1] obtained via acylcarnitine family transferase.[2]

Application

Octanoyl-L-carnitine may be used as an analytical standard in the following:
  • Separation and identification of underivatized octanoyl-L-carnitine in human plasma samples using high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS).[3]
  • Quantification of the analyte in biological samples of patients treated with continuous ambulatory peritoneal dialysis (CAPD) or automated peritoneal dialysis (APD) using high-performance liquid chromatography/tandem quadrupole mass spectrometry (HPLC-TQMS). The feasibility of adding L-carnitine to the peritoneal dialysis (PD) solution in APD-treated end-stage renal disease patients is investigated.[2]

It may be used as a test material for investigating its rate of oxidation by mitochondria, isolated from the livers of fed, starved and glucagon treated virgin and lactating rats.[4]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of glucagon treatment and starvation of virgin and lactating rats on the rates of oxidation of octanoyl-L-carnitine and octanoate by isolated liver mitochondria
Zammit AV
The Biochemical Journal, 190(2), 293-300 (1980)
L-Carnitine moiety assay: an up-to-date reappraisal covering the commonest methods for various applications
Marzo A and Curti S
Journal of Chromatography. B, Biomedical Sciences and Applications, 702(1-2), 1-20 (1997)
L-Carnitine status in end-stage renal disease patients on automated peritoneal dialysis
Liberato DL , et al.
Journal of Nephrology, 27(6), 699-706 (2014)
Minzhi Peng et al.
Journal of chromatography. A, 1319, 97-106 (2013-10-31)
A simple HPLC-MS/MS method has been established to separate and identify underivatized acylcarnitine isomers. Human plasma samples were deproteinized and concentrated. Acylcarnitines were separated on a reverse phase column and detected with triple quadrupole linear ion trap mass spectrometry. Deuterium-labeled

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service