Skip to Content
Merck
All Photos(1)

Documents

42602

Sigma-Aldrich

Propionyl-L-carnitine

≥94.0% (HPLC), suitable for LC/MS

Synonym(s):

(2R)-3-Carboxy-N,N,N-trimethyl-2-(1-oxopropoxy)-1-propanaminium inner salt, L-Carnitine propionyl ester, C3-Carnitine, Propanoyl-L-carnitine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19NO4
CAS Number:
Molecular Weight:
217.26
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

Propionyl-L-carnitine, ≥94.0% (HPLC)

Quality Level

Assay

≥94.0% (HPLC)

form

powder or crystals

optical activity

[α]/D -23±3°, c = 1 in H2O

technique(s)

LC/MS: suitable

impurities

≤10% water (calcd. from elemental analysis)

color

white to off-white

storage temp.

2-8°C

SMILES string

[O-]C(C[C@@H](OC(CC)=O)C[N+](C)(C)C)=O

InChI

1S/C10H19NO4/c1-5-10(14)15-8(6-9(12)13)7-11(2,3)4/h8H,5-7H2,1-4H3/t8-/m1/s1

InChI key

UFAHZIUFPNSHSL-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propionyl-L-carnitine (PLC) is a short-chain fatty acid ester derivative of carnitine.

Application

Propionyl-L-carnitine has been used as a standard in liquid chromatography-tandem mass spectrometry (LC-MS/MS).

Biochem/physiol Actions

Propionyl-L-carnitine (PLC) plays a key role in lipid and carbohydrate metabolism. It provides substrates to the ischemic muscle for energy production and ATP generation. PLC has the potential to treat severe peripheral arterial disease and chronic heart failure. It is present in high abundance in the urine of patients with methylmalonyl-CoA mutase (MUT) deficiency, together with methylmalonic acid. MUT is a mitochondrial enzyme that catalyzes the isomerization of methylmalonyl-CoA to succinyl-CoA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William R Hiatt et al.
Journal of cardiopulmonary rehabilitation and prevention, 31(2), 125-132 (2010-09-24)
Exercise training is established for the treatment of peripheral artery disease; however the additional benefit of pharmacologic therapy with exercise has not been studied. This trial tested the hypothesis that propionyl-L-carnitine (PLC), in combination with monitored home-based exercise training, would
Carmen Mingorance et al.
Nutrition reviews, 69(5), 279-290 (2011-04-28)
Propionyl-L-carnitine (PLC) is a naturally occurring derivative of carnitine that plays an important role in the metabolism of both carbohydrates and lipids, leading to an increase of ATP generation. PLC, however, is not only a metabolic drug; it is also
M J Cipolla et al.
Journal of vascular surgery, 29(6), 1097-1103 (1999-06-09)
The vasoactive effects of propionyl-L-carnitine (PLC) on human arteries, including endothelial and smooth muscle cell influences, were studied. Small (less than 200 microm) subcutaneous fat arteries (n = 19), obtained from human patients undergoing vascular surgery, were dissected and mounted
Santica M Marcovina et al.
Translational research : the journal of laboratory and clinical medicine, 161(2), 73-84 (2012-11-10)
Mitochondria play important roles in human physiological processes, and therefore, their dysfunction can lead to a constellation of metabolic and nonmetabolic abnormalities such as a defect in mitochondrial gene expression, imbalance in fuel and energy homeostasis, impairment in oxidative phosphorylation
Qixiao Jiang et al.
Environmental toxicology and pharmacology, 48, 116-124 (2016-10-25)
Perfluorooctanoic acid (PFOA), a persistent organic pollutant, is associated with developmental toxicity. This study investigated the mechanism of PFOA-induced developmental cardiotoxicity in chicken embryo, focusing on the interactions between developmental exposure to PFOA and the levels of l-carnitine (LC), acetyl-l-carnitine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service